Skip to Content
Merck
All Photos(1)

Key Documents

1.10213

Millipore

Tryptose

suitable for microbiology

Synonym(s):

Tryptose, Tryptic digested protein

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106212
NACRES:
NA.74
Pricing and availability is not currently available.

biological source

animal

Quality Level

Assay

11.0-13.0% amino-nitrogen basis (purity)

form

solid

packaging

pkg of 1 kg

storage condition

protect from light (Store dry and tightly closed)

color

light brown

pH

6-7 (20 °C, 20 g/L in H2O)

solubility

560 g/L

bulk density

300 kg/m3

application(s)

microbiology

storage temp.

15-25°C

General description

Tryptose is a mixture of tryptic-digested proteins. It is a suitable nutrient base material for applications in the biotechnology and pharmaceutical industry.It is recommended for use in media for the cultivation of a wide range of fastidious bacteria such as Brucella, Streptococcoccus, Pneumococcus, Meningococcus spp., etc

Application

Tryptose is used in cell culture, molecular genetic media, and as a nutritive substrate for the cultivation and resuscitation of a large variety of fastidious microorganisms.

Analysis Note

Appearance: Brownishyellowfine powder
Suitability for microbiology: passes test
pH-value (5 % solution): 6.5 - 7.5
Total nitrogen (N): 11.0 - 13.0 %
Amino nitrogen: 3.0 - 5.0 %
Sulfated ash: ≤ 15.0 %

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service