Skip to Content
Merck
All Photos(1)

Key Documents

1.10141

Millipore

Salmosyst Selective Supplement

for Salmonella spp., pack of 250 tablets, for use with Salmosyst Broth Base

Synonym(s):

Salmonella selective supplement

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106212
NACRES:
NA.74
Pricing and availability is not currently available.

form

solid

Quality Level

composition

Brilliant green, 0.0007 g (per tablet)
Calcium carbonate, 0.1 g (per tablet)
Ox bile, 0.08 g (per tablet)
Potassium tetrathionate, 0.2 g (per tablet)

packaging

pack of 250 tablets

technique(s)

microbiological culture: suitable

pH

6.9-7.3 (20 °C, 10 g/L in H2O)

application(s)

food and beverages
microbiology

compatibility

for use with Salmosyst Broth Base

storage temp.

15-25°C

suitability

Salmonella spp.

General description

Salmosyst Broth Base enriches all the microorganisms present in the sample material during non-selective pre-enrichment. The addition of the selective reagents in the form of Salmosyst Selective Supplement inhibits the growth of the accompanying organisms, except Salmonella spp.

Application

Salmosyst Selective Supplement is an additive for the preparation of Salmosyst Broth Base. It is used for the two-step enrichment of sub-lethally injured Salmonella spp. from foodstuffs and feeds.

Analysis Note

Disintegration time (25 °C): 4 - 10 min
Growth test:
passes test
: conforms

Legal Information

related product

Product No.
Description
Pricing

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service