Skip to Content
Merck
All Photos(1)

Key Documents

1.09877

Millipore

OGYE selective supplement

for yeasts, for molds, pkg of 10 vials, for use with OGYE agar, Base

Synonym(s):

Oxytetracyclin-Glucose-Yeast Extract selective supplement

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106212
NACRES:
NA.74
Pricing and availability is not currently available.

composition

Oxytetracycline, 0.05 g (in a buffered base )

Quality Level

packaging

pkg of 10 vials

technique(s)

microbiological culture: suitable

application(s)

food and beverages
microbiology

compatibility

for use with OGYE agar, Base

storage temp.

2-8°C

suitability

molds
yeasts

General description

OGYE Selective Supplement contains oxytetracycline in lyophilized form. It suppresses the growth of accompanying bacterial flora during culturing yeasts and molds.

Application

OGYE Selective Supplement is an additive used during the OGYE Agar Base during the preparation of OGYE Selective Agar for enumeration of yeasts and molds in food.

Compatibility

For use with OGYE agar, Base (1.05978)

Linkage

Replaces: 1.09877.0017

Analysis Note

Tested in OGYE-agar base art. no. 1.05978.
Recovery on test medium
- Candida albicans ATCC 10231 (WDCM 00054):
≥ 50 %
- Saccharomyces cerevisiae ATCC 9763 (WDCM 00058): ≥ 50 %
- Aspergillus brasiliensis (formerly A. niger) ATCC 16404 (WDCM 00053): ≥ 50 %
- Penicillium aurantiogriseum ATCC 16025: ≥ 50 %
Growth
- Bacillus subtilis ATCC 6633 (WDCM 00003):
none
- Escherichia coli ATCC 25922 (WDCM 00013): none
Incubation: 3 up to 5 days; 28 °C; aerobic.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service