Skip to Content
Merck
All Photos(1)

Key Documents

1.09222

Millipore

MAS-100 Iso NT® Perforated Lid

stainless steel, no. of holes, 400 hole installed ( ), hole diam. 0.7 mm

Synonym(s):

collection head, sieve head

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122409
NACRES:
NA.74
Pricing and availability is not currently available.

material

stainless steel

Agency

non-compendial

feature

no. of holes 400 hole installed ( )

packaging

pkg of 1 unit

parameter

11 m/s impaction speed (at flow rate of 100 L/min)

hole diam.

0.7 mm

application(s)

air monitoring
pharmaceutical

compatibility

for use with 90mm Petri Dishes
for use with MAS-100®

storage temp.

no temp limit

General description

MAS-100 ISO NT® Perforated Lid is made from stainless steel and can be used with MAS-100 ISO NT® and MAS-100 ISO MH® microbial air samplers. It is compatible with the standard and easy clean base. The sieve of the perforated lid is the core of the air sampler. Each hole must have an identical diameter and length to accelerate the aspirated air correctly. The 3mm thickness of the perforated sieve guarantee a well-defined impaction jet. The impaction speed defines the physical sampling efficiency. The cut-off value D50 (the particle size at which 50% of the particles impact in the agar) must be identical between holes and between lids. That is why intense quality control of the sieves is performed. The sieve pattern with 400 holes of a diameter of 0.7 mm provides an impaction speed of approximately 11 m/s at a flow rate of 100 SLPM. This impaction speed allows a high physical efficiency, meaning a sampling efficiency of more the 50% of small particles at a size of less than 1.62 μm (D50, calculated) while maintaining the viability of microorganisms during impaction on the agar surface. The edge protection prevents scratching of the sealing surface which would lead to aspirating air through the gap. The perforated lid can be autoclaved and disinfected with all disinfectants compatible to stainless steel in order to avoid false positive results in high grade cleanrooms.

Application

MAS-100 ISO NT® Perforated Lid, Stainless Steel 400-hole edge protected is used for microbial monitoring of ambient air with MAS-100 Iso family microbial air samplers in isolators, RABS and high-grade cleanrooms. Microbial air sampling devices are extensively used in Pharmaceutical applications.

Legal Information

MAS-100 is a registered trademark of MBV AG
MAS-100 ISO MH is a registered trademark of MBV AG
MAS-100 ISO NT is a registered trademark of MBV AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service