Skip to Content
Merck
All Photos(1)

Key Documents

1.09218

Sigma-Aldrich

Gram′s crystal violet solution

for the Gram staining method

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116130
NACRES:
NA.74
Pricing and availability is not currently available.

Quality Level

form

liquid

IVD

for in vitro diagnostic use

technique(s)

microbe id | staining: suitable

transition temp

flash point 47 °C

density

0.99 g/cm3 at 20 °C

application(s)

clinical testing
diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

General description

Gram′s crystal violet solution for the Gram staining method, is a ready-to-use solution used in bacteriology. The Gram staining allows a fast differentiation of bacteria in Gram-positive and Gram-negative.The mureine structure of the bacteria wall is the basis of the color affinity. In the first step, bacteria will be stained with Gram′s crystal violet solution (Product number 1.09218), an aniline dye. After the treatment with iodine solution (Lugol′s solution stabilized with PVP, Product number 1.00567), a dye-iodine complex will form. During the decolorizing step (Gram′s decolorizing solution, Product number 1.10218), this complex stays in the multilayer mureine structure of the cell wall of Gram-positive bacteria - they will appear blue-violet. Gram-negative bacteria, by contrast, have a cell wall consisting of a single-layered murein structure, and correspondingly re-release the staining dye with the decoloring solution. Gram-negative bacteria will be counterstained with safranine solution (Gram′s safranine solution (Product number 1.09217) and will then appear pink to red.
A bottle with 500 ml Gram′s crystal violet solution is sufficient for staining up to 250 slides. This product is registered as IVD and CE marked. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Analysis Note

Suitability for microscopy: passes test
gram-positve microorganisms: blue-violet
gram-negative microorganisms: pink to red

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

116.6 °F

Flash Point(C)

47 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service