Skip to Content
Merck
All Photos(2)

Documents

1.08883

Supelco

Zinc sulfate heptahydrate

for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Synonym(s):

Zinc

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZnSO4 · 7H2O
CAS Number:
Molecular Weight:
287.56
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-793-3
NACRES:
NA.21

grade

ACS reagent

Quality Level

Agency

reag. ISO
reag. Ph. Eur.
suitable for SM 5210

product line

EMSURE®

Assay

99.5-103.0% (complexometric)

form

solid

potency

1260 mg/kg LD50, oral (Rat)

impurities

≤0.01% Insoluble matter

pH

4-6 (20 °C, 50 g/L in H2O)

mp

100 °C

solubility

965 g/L

density

1.97 g/cm3 at 20 °C

bulk density

800‑1000 kg/m3

anion traces

chloride (Cl-): ≤0.0005%
nitrate (NO3-): ≤0.002%

cation traces

As: ≤0.00005%
Ca: ≤0.001%
Cd: ≤0.0005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.001%
Mg: ≤0.001%
Mn: ≤0.0003%
N: ≤0.0005%
NH4+: ≤0.001%
Na: ≤0.001%
Pb: ≤0.001%

storage temp.

2-30°C

SMILES string

O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O

InChI

1S/H2O4S.7H2O.Zn/c1-5(2,3)4;;;;;;;;/h(H2,1,2,3,4);7*1H2;/q;;;;;;;;+2/p-2

InChI key

RZLVQBNCHSJZPX-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Validation of a simple liquid chromatography coupled to tandem mass spectrometry method for the simultaneous determination of tacrolimus, sirolimus, everolimus and cyclosporin A in dried matrix on paper discs.: This study validates a liquid chromatography-mass spectrometry method enhanced by zinc sulfate heptahydrate for analyzing immunosuppressant drugs. This development represents a significant advance in clinical diagnostics, providing faster and more reliable drug monitoring (Bressán et al., 2021).

  • Adduct simplification in the analysis of cyanobacterial toxins by matrix-assisted laser desorption/ionization mass spectrometry.: Research utilizing zinc sulfate heptahydrate in matrix-assisted laser desorption/ionization (MALDI) mass spectrometry to simplify the analysis of cyanobacterial toxins. This study enhances the understanding of environmental toxins, showcasing the role of zinc sulfate heptahydrate in improving analytical accuracy and efficiency (Howard et al., 2007).

Linkage

Replaces: ZX0105; ZX0105-1; ZX0105-20

Analysis Note

Assay (complexometric): 99.5 - 103.0 %
Identity: passes test
Appearance of solution: passes test
Insoluble matter: ≤ 0.01 %
pH-value (5 %; water, 25 °C): 4.4 - 5.6
Chloride (Cl): ≤ 0.0005 %
Total nitrogen (N): ≤ 0.0005 %
Nitrate (NO₃): ≤ 0.002 %
As (Arsenic): ≤ 0.00005 %
Ca (Calcium): ≤ 0.001 %
Cd (Cadmium): ≤ 0.0005 %
Cu (Copper): ≤ 0.0005 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.001 %
Mg (Magnesium): ≤ 0.001 %
Mn (Manganese): ≤ 0.0003 %
Na (Sodium): ≤ 0.001 %
NH₄ (Ammonium): ≤ 0.001 %
Pb (Lead): ≤ 0.001 %
Corresponds to ACS, ISO, Reag. Ph Eur

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service