Skip to Content
Merck
All Photos(1)

Documents

1.08430

Supelco

Indicator buffer tablets

for measuring water hardness with the Titriplex®system

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161703
NACRES:
NB.21

product name

Indicator buffer tablets, for measuring water hardness with the Titriplex® system

form

solid

reaction suitability

reaction type: volumetric

technique(s)

titration: suitable

weight

≥0.20 g

storage temp.

no temp limit

General description

for the determination of water hardness with Titriplex® solutions

Analysis Note

Weight (min): ≥ 0.20 g
Weight (max): ≤ 0.23 g
Disintegration time (at 25 °C): ≤ 150 s
Application test: conforms

Legal Information

Titriplex is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service