Skip to Content
Merck
All Photos(1)

Key Documents

1.08417

Supelco

Titriplex® II

for analysis (Ethylenedinitrilotetraacetic acid, EDTA), ACS, reag. Ph. Eur.

Synonym(s):

Titriplex® II, EDTA, Edetic acid, Diaminoethane-tetraacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N2O8
CAS Number:
Molecular Weight:
292.24
MDL number:
UNSPSC Code:
12352106
EC Index Number:
200-449-4
NACRES:
NA.21
Pricing and availability is not currently available.

Product Name

Titriplex® II, for analysis (ethylenedinitrilotetraacetic acid) ACS,Reag. Ph Eur

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

vapor pressure

<0.013 hPa ( 20 °C)

product line

Titripur®

Assay

≥99.4-100.6% (complexometric)

form

solid

autoignition temp.

>200 °C

potency

2580 mg/kg LD50, oral (Rat)

reaction suitability

reaction type: Complexometric reactions

technique(s)

titration: suitable

pH

2.5 (23 °C, 10 g/L in H2O, slurry)

mp

220 °C (decomposition)

transition temp

flash point >400 °C (DIN 51758)

solubility

0.5 g/L

density

0.86 g/cm3 at 20 °C

bulk density

600 kg/m3

cation traces

Ca: ≤10 ppm
Fe: ≤10 ppm
Mg: ≤5 ppm
heavy metals (as Pb): ≤10 ppm

storage temp.

15-25°C

SMILES string

N(CCN(CC(=O)O)CC(=O)O)(CC(=O)O)CC(=O)O

InChI

1S/C10H16N2O8/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20)

InChI key

KCXVZYZYPLLWCC-UHFFFAOYSA-N

Analysis Note

Assay (complexometric): 99.4 - 100.6 %
Identity (IR-spectrum): conforms
Appearance: White to almost white, fine crystalline to crystalline powder.
in ammonia insoluble components: ≤ 0.005 %
Heavy metals (as Pb): ≤ 10 ppm
Ca (Calcium): ≤ 10 ppm
Fe (Iron): ≤ 10 ppm
Mg (Magnesium): ≤ 5 ppm
Nitrilotriacetic acid: ≤ 0.1 %
Sulfated ash: ≤ 0.1 %
Water (according to Karl Fischer): ≤ 0.2 %
ACS and Reag. Ph Eur conforms

Legal Information

Titriplex is a registered trademark of Merck KGaA, Darmstadt, Germany
Titripur is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Validated HPLC method (based on Ph. Eur. Monograph 2668) for quantifying catechins and caffeine in decaffeinated green tea using a Chromolith® High Resolution RP-18e column with advantages in terms of backpressure and matrix robustness.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service