Skip to Content
Merck

1.08307

SAFC

Tris(hydroxymethyl)aminomethane

(Trometamol) high purity, EMPROVE® EXPERT, Ph. Eur., BP, ChP, JPC, USP

Pharma Manufacturing

Synonym(s):

Aminomethylidine trimethanol, 2-Amino-2-hydroxymethyl-1.3-propanediol, TRIS-buffer, Tromethamine, Tris(Hydroxymethyl)aminomethane, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris base, Trometamol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2C(CH2OH)3
CAS Number:
Molecular Weight:
121.14
Beilstein:
741883
MDL number:
UNSPSC Code:
12352209
EC Index Number:
201-064-4
Pricing and availability is not currently available.

Agency

BP
ChP
JPC
Ph. Eur.
USP

Quality Level

vapor pressure

0.0267 hPa ( 20 °C)

description

EMPROVE® EXPERT Ph Eur,BP,JPC,USP,ACS

product line

EMPROVE® EXPERT

form

solid

feature

RNase and DNase free

pH

10.2-10.6 (20 °C, 6 g/L in H2O)

useful pH range

7-9

pKa (25 °C)

8.1

bp

219-220 °C/10 mmHg (lit.)
219-220 °C/13.3 hPa

mp

167-172 °C (lit.)

density

1.35 g/cm3 at 23 °C

application(s)

liquid formulation
pharmaceutical
semi-solid formulation

storage temp.

no temp limit

SMILES string

NC(CO)(CO)CO

InChI

1S/C4H11NO3/c5-4(1-6,2-7)3-8/h6-8H,1-3,5H2

InChI key

LENZDBCJOHFCAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Tris(hydroxymethyl)aminomethane (Trometamol) is a commonly used buffer component in biomolecule downstream chromatography steps as well as final liquid formulation.
Due to its low polyoxymethylene limits, Tris(hydroxymethyl)aminomethane (Trometamol) Emprove® Expert is suitable for use at elevated concentrations without risking precipitation. Also, low microbial and endotoxin limits facilitate the use of this product in high-risk applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This overview describes the challenges when developing parenteral formulations and strategies to overcome them and reduce the risk of production.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service