Skip to Content
Merck
All Photos(1)

Documents

1.08294

Supelco

Thorin

indicator for sulfate titration

Synonym(s):

Thorin, 1-(2-Arsonophenylazo)-2-hydroxy-3,6-naphthalene-disulfonic acid sodium salt, 2-(2-Hydroxy-3,6-disulfo-1-naphthylazo)-benzene-arsonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H11AsN2Na2O10S2
CAS Number:
Molecular Weight:
576.30
UNSPSC Code:
12352103
EC Index Number:
222-993-1
NACRES:
NB.21

product name

Thorin, indicator for sulfate titration

Quality Level

form

solid

impurities

≤25% Water (Karl Fischer)

pH

6.1 (20 °C, 10 g/L in H2O)

bulk density

700 kg/m3

λmax

480-490 nm (propan-2-ol)

storage temp.

2-30°C

InChI

1S/C16H13AsN2O10S2.2Na/c20-16-14(31(27,28)29)8-9-7-10(30(24,25)26)5-6-11(9)15(16)19-18-13-4-2-1-3-12(13)17(21,22)23;;/h1-8,18H,(H2,21,22,23)(H,24,25,26)(H,27,28,29);;/q;2*+1/p-2/b19-15+;;

InChI key

ZQMPNLOZJUWHSA-WTBAQZMLSA-L

Analysis Note

Identity (UV/VIS-Spectrum): passes test
Absorption maximum λmax. (Propan-2-ol): 480 - 490 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.025 g/l;propan-2-ol; calc. on anhydrous substance): 250 - 350
Water (according to Karl Fischer): ≤ 25 %
Suitability as indicator (for sulfate titration): passes test

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service