Skip to Content
Merck
All Photos(1)

Documents

1.08180

Supelco

2-Thiobarbituric acid

reagent for sorbic acid

Synonym(s):

2-Thiobarbituric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4N2O2S
CAS Number:
Molecular Weight:
144.15
MDL number:
UNSPSC Code:
12352115
EC Index Number:
207-985-8
NACRES:
NB.21

Quality Level

potency

>5000 mg/kg LD50, oral (Rat)

pH

1.7-1.9 (20 °C, 10 g/L in H2O)

mp

235 °C

bulk density

210 kg/m3

storage temp.

15-25°C

InChI

1S/C4H4N2O2S/c7-2-1-3(8)6-4(9)5-2/h1H2,(H2,5,6,7,8,9)

InChI key

RVBUGGBMJDPOST-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric; calculated on dried substance): ≥ 99
Identity (IR-spectrum): passes test
pH-value (1 %; water): 1.7 - 1.9
Loss on drying (20 mbar, 60 °C): ≤ 1
Suitability as reagent (for sorbic acid): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service