Skip to Content
Merck
All Photos(1)

Key Documents

1.07857

Supelco

Strontium chloride hexahydrate

for atomic absorption spectroscopy (max. 0.0005% Ca)

Synonym(s):

Strontium chloride hexahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cl2Sr · 6H2O
CAS Number:
Molecular Weight:
266.62
MDL number:
UNSPSC Code:
12352302
EC Index Number:
233-971-6
grade:
for inorganic trace analysis
form:
solid
solubility:
2058 g/L
Pricing and availability is not currently available.

grade

for inorganic trace analysis

Quality Level

form

solid

pH

5-7 (25 °C, 50 g/L in H2O)

mp

61 °C

solubility

2058 g/L

density

1.95 g/cm3 at 20 °C

bulk density

1050 kg/m3

storage temp.

2-30°C

InChI

1S/2ClH.6H2O.Sr/h2*1H;6*1H2;/q;;;;;;;;+2/p-2

InChI key

AMGRXJSJSONEEG-UHFFFAOYSA-L

Analysis Note

Assay (complexometric): ≥ 99.0 %
Ba (Barium): ≤ 0.01 %
Ca (Calcium): ≤ 0.0005 %
Cu (Copper): ≤ 0.0002 %
Fe (Iron): ≤ 0.0005 %
K (Potassium): ≤ 0.01 %
Mg (Magnesium): ≤ 0.0002 %
Na (Sodium): ≤ 0.01 %
Pb (Lead): ≤ 0.0002 %
Zn (Zinc): ≤ 0.0002 %

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service