Skip to Content
Merck
All Photos(2)

Documents

1.07758

Millipore

D(-)-Sorbitol

extra pure, suitable for microbiology

Synonym(s):

D-Sorbitol, D-Glucitol, D-Glucitol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14O6
CAS Number:
Molecular Weight:
182.17
Beilstein:
1721899
MDL number:
UNSPSC Code:
41106212
EC Index Number:
200-061-5
NACRES:
NA.74

vapor density

<1 (vs air)

Quality Level

vapor pressure

<0.1 mmHg ( 25 °C)

form

solid

autoignition temp.

420 °C

potency

15900 mg/kg LD50, oral (Rat)

pH

6-7 (20 °C, 100 g/L in H2O)

mp

98-100 °C (lit.)

transition temp

flash point >100 °C

solubility

2350 g/L

density

1.49 g/cm3 at 20 °C

bulk density

450 kg/m3

application(s)

microbiology

storage temp.

15-25°C

SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)CO

InChI

1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2/t3-,4+,5-,6-/m1/s1

InChI key

FBPFZTCFMRRESA-JGWLITMVSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Identity: passes test
Chloride (Cl): ≤ 0.002 %
Sulfate (SO₄): ≤ 0.01
Water: ≤ 1.0 %
Suitability for microbiology: passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service