Skip to Content
Merck
All Photos(2)

Documents

1.07736

Supelco

Silica gel 60 H

for thin-layer chromatography

Synonym(s):

Silica gel

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41115711
EC Index Number:
231-545-4
NACRES:
NB.21

material

silica gel 60 matrix

Quality Level

form

powder

potency

>5000 mg/kg LD50, skin (Rabbit)

feature

binder: no
fluorescent indicator: no

packaging

pkg of 25 kg

technique(s)

thin layer chromatography (TLC): suitable

particle size

5-40 μm

pore size

60 Å medium pore diameter

pH

7 (20 °C, 100 g/L in H2O, slurry)

bp

2230 °C

mp

>1600 °C

bulk density

200‑800 kg/m3

storage temp.

no temp limit

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Standardized sorbents ensure reliable results every time. Our broad spectrum meets all kinds of needs. In TLC, silica gel 60 sorbent is considered the most versatile. Our assortment of silica gel 60 features different grades with particle sizes ranging from 5-40 µm: silica with a gypsum binder, silica with no foreign binder, and silica gel with a fluorescence indicator. We also offer high-quality aluminum oxide, cellulose microcrystalline, and kieselgur (also known as diatomaceous earth).

Application


  • Exploring the versatility of sulfur-containing heterocyclic metal complexes: Application in medical and prospects of visible-light-driven photocatalysis: Investigates the medical and environmental applications of sulfur-containing metal complexes, showing potential in light-driven processes and therapeutic areas (Daisylet et al., 2024).

  • PAHs removal by soil washing with thiacalix[4]arene tetrasulfonate: Describes the use of sulfur-based compounds in environmental cleanup processes, particularly for the removal of persistent organic pollutants in soil (Xing et al., 2024).



Linkage

Replaces: 7736-9; 7736

Analysis Note

pH-value (10% suspension): 6.5 - 7.5
Fe (Iron): ≤ 0.02%
Particle size: 90% <55 µm
Application test: passes test
The data to the particle size are related to the pure Silica gel before additive addition.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service