Skip to Content
Merck
All Photos(1)

Key Documents

1.07709

Millipore

Selenite cystine enrichment broth

Synonym(s):

Selenite-cystine broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
Pricing and availability is not currently available.

form

medium granules

technique(s)

microbiological culture: suitable

pH

6.8-7.2 (37 °C, 23 g/L in H2O)

solubility

23 g/L

application(s)

clinical testing
food and beverages
microbiology
pathogen testing

General description

for the enrichment of salmonellae

Storage and Stability

Store below +15°C.

Analysis Note

Appearance (clearness):clear
Appearance (colour):yellowish
pH-value (25 °C):6.8 - 7.2
Inoculum on reference medium (Salmonella typhimurium ATCC 14028 (WDCM 00031)):10 - 100
Inoculum on reference medium (Escherichia coli ATCC 25922 (WDCM 00013)):≥ 1E+04
Growth (Salmonella typhimurium ATCC 14028 (WDCM 00031)):+
Growth (Escherichia coli ATCC 25922 (WDCM 00013)):inhibited
Enrichment from a mixed inoculum after 24 hours (Salmonella typhimurium ATCC 14028 (WDCM 00031)):+
Enrichment from a mixed inoculum after 24 hours (Escherichia coli ATCC 25922 (WDCM 00013)):-
Incubation: 24 h at 35 °C; Detection of Salmonellae from the mixed culture with XLD agar

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Supplementary Hazards

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service