Skip to Content
Merck
All Photos(2)

Documents

1.07026

Supelco

Diethyl ether

≥99.5% (GC), EMPARTA® ACS reagent, contains 5-10 ppm 2, 6-Di-tert-butyl-4-methylphenol (BHT) as stabilizer

Synonym(s):

Et2O, Ethoxyethane, Ethyl ether, Ether, Ether, Ethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2)2O
CAS Number:
Molecular Weight:
74.12
Beilstein:
1696894
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-467-2
NACRES:
NA.05

product name

Diethyl ether, for analysis EMPARTA® ACS

grade

ACS reagent

Quality Level

vapor density

2.6 (vs air)

vapor pressure

587 hPa ( 20 °C)

product line

EMPARTA®

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

320 °F

potency

1215 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

contains

5-10 ppm 2,6-Di-tert-butyl-4-methylphenol (BHT) as stabilizer

expl. lim.

36.5 %

impurities

≤0.000015% Peroxide (as H2O2)
≤0.0002 meq/g Titrable acid
≤0.001% Carbonyl compounds (as HCHO)
≤0.1% Water

evapn. residue

≤0.001%

color

clearAPHA: ≤10

refractive index

n20/D 1.3530 (lit.)

bp

34.6 °C (lit.)

mp

−116 °C (lit.)

transition temp

flash point -40 °C

density

0.706 g/mL at 25 °C (lit.)

storage temp.

15-25°C

SMILES string

CCOCC

InChI

1S/C4H10O/c1-3-5-4-2/h3-4H2,1-2H3

InChI key

RTZKZFJDLAIYFH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Inhibition of Food-Borne Pathogen Growth and Biogenic Amine Synthesis by Spice Extracts.: This research explores the efficacy of spice extracts in inhibiting pathogen growth, with diethyl ether used in the extraction process to isolate active compounds (Kuley et al., 2024).


  • β-sitosterol isolated from the leaves of Trema orientalis (Cannabaceae) promotes viability and proliferation of BF-2 cells.: Features the use of diethyl ether in the extraction of β-sitosterol, showcasing its application in bioactive compound isolation for medical research (Mekarunothai et al., 2024).


  • Solvent Extraction of PDMS Tubing as a New Method for the Capture of Volatile Organic Compounds from Headspace.: Introduces a novel method for capturing volatile organic compounds using diethyl ether, enhancing analytical capabilities in chemical ecology (Thomas et al., 2024).


  • Metal Vapour Synthesis of an Organometallic Barium(0) Synthon.: Describes the critical role of diethyl ether in the synthesis of complex organometallic compounds, underlining its utility in advanced materials science (Townrow et al., 2024).


Analysis Note

Purity (GC): ≥ 99.5 %
Identity (IR): conforms
Appearance: clear
Color: ≤ 10 Hazen
Titrable acid: ≤ 0.0002 meq/g
Boiling point: 34 - 35 °C
Peroxide (as H₂O₂): ≤ 0.000015 %
Carbonyl compounds (as HCHO): ≤ 0.001 %
Evaporation residue: ≤ 0.001 %
Water: ≤ 0.1 %
The product is stabilized with 5-10 ppm 2,6-Di-tert-butyl-4-methylphenol (BHT).

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 1 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-40.0 °F - closed cup

Flash Point(C)

-40 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service