Skip to Content
Merck
All Photos(1)

Key Documents

1.06546

Supelco

Sodium nitrate

99.99 Suprapur®

Synonym(s):

Sodium nitrate, Nitric acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
NNaO3
CAS Number:
Molecular Weight:
84.99
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-554-3
NACRES:
NA.21
Assay:
99.99%
grade:
for inorganic trace analysis
form:
solid
solubility:
874 g/L
Pricing and availability is not currently available.

grade

for inorganic trace analysis

Quality Level

Assay

99.99%

form

solid

potency

1267 mg/kg LD50, oral (Rat)

pH

5.5-8.0 (20 °C, 50 g/L in H2O)

mp

308 °C

solubility

874 g/L

density

2.26 g/cm3 at 20 °C

bulk density

1200 kg/m3

storage temp.

2-30°C

SMILES string

[Na+].[N+](=O)([O-])[O-]

InChI

1S/NO3.Na/c2-1(3)4;/q-1;+1

InChI key

VWDWKYIASSYTQR-UHFFFAOYSA-N

Application


  • Sodium salt promoted the generation of nano zero valent iron by carbothermal reduction: For activating peroxydisulfate to degrade antibiotic.: Research demonstrates the use of sodium nitrate in the synthesis of nano zero valent iron, which is effective in activating peroxydisulfate for the degradation of antibiotics in wastewater treatment, offering a potential solution to antibiotic pollution (Xue C et al., 2024).


Analysis Note

Purity (metallic): ≥ 99.99 %
Assay (acidimetric): ≥ 99 %
Chloride (Cl): ≤ 5 ppm
Iodate (IO₃): ≤ 5 ppm
Nitrite (NO₂): ≤ 10 ppm
Phosphate (PO₄): ≤ 2 ppm
Sulfate (SO₄): ≤ 30 ppm
Al (Aluminium): ≤ 0.05 ppm
Ba (Barium): ≤ 5.0 ppm
Ca (Calcium): ≤ 0.10 ppm
Cd (Cadmium): ≤ 0.010 ppm
Ce (Cerium): ≤ 0.010 ppm
Co (Cobalt): ≤ 0.010 ppm
Cr (Chromium): ≤ 0.010 ppm
Cs (Cesium): ≤ 5 ppm
Cu (Copper): ≤ 0.010 ppm
Eu (Europium): ≤ 0.010 ppm
Fe (Iron): ≤ 0.05 ppm
K (Potassium): ≤ 5.0 ppm
La (Lanthanum): ≤ 0.010 ppm
Mg (Magnesium): ≤ 0.05 ppm
Mn (Manganese): ≤ 0.010 ppm
Ni (Nickel): ≤ 0.010 ppm
Pb (Lead): ≤ 0.010 ppm
Rb (Rubidium): ≤ 5.0 ppm
Sc (Scandium): ≤ 0.010 ppm
Sm (Samarium): ≤ 0.010 ppm
Sr (Strontium): ≤ 0.10 ppm
Tl (Thallium): ≤ 0.01 ppm
Y (Yttrium): ≤ 0.010 ppm
Yb (Ytterbium): ≤ 0.010 ppm
Zn (Zinc): ≤ 0.010 ppm

Legal Information

SUPRAPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service