Skip to Content
Merck
All Photos(1)

Key Documents

1.06053

Supelco

Diiodomethane

for the separation of minerals

Synonym(s):

Diiodomethane, Methylene iodide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH2I2
CAS Number:
Molecular Weight:
267.84
MDL number:
UNSPSC Code:
12352101
EC Index Number:
200-841-5
NACRES:
NB.21
Assay:
≥98.5% (GC)
bp:
181 °C/1013 hPa (decomposition)
Pricing and availability is not currently available.

Quality Level

Assay

≥98.5% (GC)

form

liquid

potency

76 mg/kg LD50, oral (Rat)

bp

181 °C/1013 hPa (decomposition)

mp

6 °C

solubility

0.8 g/L

density

3.3 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

ICI

InChI

1S/CH2I2/c2-1-3/h1H2

InChI key

NZZFYRREKKOMAT-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.5 % (a/a)
Density (d 20 °C/ 4 °C): 3.310 - 3.320
Water (K. F.): ≤ 0.02 %
Free acid (as HI): ≤ 0.005 %
Evaporation residue: ≤ 0.005 %
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service