Skip to Content
Merck
All Photos(1)

Key Documents

1.05686

Supelco

Lithium fluoride

for inorganic trace analysis, 99.99 Suprapur®

Synonym(s):

Lithium fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
FLi
CAS Number:
Molecular Weight:
25.94
MDL number:
UNSPSC Code:
12352302
EC Index Number:
232-152-0
NACRES:
NA.21
Pricing and availability is not currently available.

grade

for inorganic trace analysis

Quality Level

Assay

99.99%

form

solid

potency

143 mg/kg LD50, oral (Rat)

bp

1676 °C/1013 hPa

mp

870 °C

density

2.64 g/cm3 at 20 °C

bulk density

600 kg/m3

storage temp.

2-30°C

SMILES string

[Li+].[F-]

InChI

1S/FH.Li/h1H;/q;+1/p-1

InChI key

PQXKHYXIUOZZFA-UHFFFAOYSA-M

Related Categories

Application


  • Radical Polymer-based Positive Electrodes for Dual-Ion Batteries: Describes the enhancement of battery performance through the use of γ-Butyrolactone-based electrolytes in conjunction with radical polymer-based positive electrodes, highlighting advances in lithium carbonate applications in battery technology (Rudolf K, et al., 2024).

  • Analysis of Trace Impurities in Lithium Carbonate: Focuses on the precision analysis of impurities within pharmaceutical grade lithium carbonate, crucial for maintaining high purity standards in pharmaceutical applications (Suárez A, et al., 2024).

  • Effects of Central Metal Ion on Binuclear Metal Phthalocyanine-Based Redox Mediator for Lithium Carbonate Decomposition: Investigates the role of central metal ions in enhancing the efficiency of lithium carbonate decomposition, important for environmental lithium analysis and recovery (Yan Q, et al., 2024).

  • Selective preparation of lithium carbonate from overhaul slag by high temperature sulfuric acid roasting - Water leaching: Details a method for extracting high-purity lithium carbonate from industrial waste, demonstrating an environmentally friendly approach to lithium recovery (Dong L, et al., 2024).

  • Reactive molecular dynamics simulations of lithium-ion battery electrolyte degradation: Explores the degradation mechanisms of lithium-ion battery electrolytes using simulations, contributing to the development of more durable lithium battery materials (Mabrouk Y, et al., 2024).

Analysis Note

Purity (metallic): ≥ 99.99 %
Ba (Barium): ≤ 5.0 ppm
Ca (Calcium): ≤ 2.0 ppm
Cd (Cadmium): ≤ 0.5 ppm
Co (Cobalt): ≤ 0.5 ppm
Cs (Cesium): ≤ 20 ppm
Cu (Copper): ≤ 0.5 ppm
Fe (Iron): ≤ 0.5 ppm
K (Potassium): ≤ 10 ppm
Mg (Magnesium): ≤ 0.5 ppm
Mn (Manganese): ≤ 0.5 ppm
Na (Sodium): ≤ 10 ppm
Ni (Nickel): ≤ 0.5 ppm
Pb (Lead): ≤ 0.5 ppm
Rb (Rubidium): ≤ 10 ppm
Sr (Strontium): ≤ 5 ppm
Zn (Zinc): ≤ 0.5 ppm

Legal Information

SUPRAPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service