Skip to Content
Merck
All Photos(2)

Documents

1.05044

Supelco

Potassium iodide

99.995 Suprapur®

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KI
CAS Number:
Molecular Weight:
166.00
MDL number:
UNSPSC Code:
12352302
EC Index Number:
231-659-4
NACRES:
NB.21

grade

for inorganic trace analysis

Quality Level

vapor pressure

1 mmHg ( 745 °C)

Assay

99.995%

form

solid

potency

2779 mg/kg LD50, oral (Rat)

pH

6.9 (20 °C, 50 g/L in H2O)

bp

1325 °C/1013 hPa

mp

681 °C (lit.)

solubility

1430 g/L

bulk density

1500 kg/m3

storage temp.

2-30°C

SMILES string

[K+].[I-]

InChI

1S/HI.K/h1H;/q;+1/p-1

InChI key

NLKNQRATVPKPDG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Analysis Note

Purity (metallic): ≥ 99.995 %
Assay (argentometric): ≥ 99.5 %
Chloride and Bromide (as Cl): ≤ 100 ppm
Phosphate (PO₄): ≤ 10 ppm
Sulfate (SO₄): ≤ 10 ppm
Total nitrogen (N): ≤ 10.0 ppm
Al (Aluminium): ≤ 0.010 ppm
Ba (Barium): ≤ 5.0 ppm
Ca (Calcium): ≤ 0.05 ppm
Cd (Cadmium): ≤ 0.010 ppm
Ce (Cerium): ≤ 0.010 ppm
Co (Cobalt): ≤ 0.010 ppm
Cr (Chromium): ≤ 0.010 ppm
Cu (Copper): ≤ 0.010 ppm
Eu (Europium): ≤ 0.010 ppm
Fe (Iron): ≤ 0.050 ppm
La (Lanthanum): ≤ 0.010 ppm
Mg (Magnesium): ≤ 0.05 ppm
Mn (Manganese): ≤ 0.010 ppm
Na (Sodium): ≤ 5 ppm
Ni (Nickel): ≤ 0.010 ppm
Pb (Lead): ≤ 0.010 ppm
Sc (Scandium): ≤ 0.010 ppm
Sm (Samarium): ≤ 0.010 ppm
Sr (Strontium): ≤ 0.10 ppm
Tl (Thallium): ≤ 0.01 ppm
Y (Yttrium): ≤ 0.010 ppm
Yb (Ytterbium): ≤ 0.010 ppm
Zn (Zinc): ≤ 0.010 ppm

Legal Information

SUPRAPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Oral

Target Organs

Thyroid

Storage Class Code

6.1D - Non-combustible, acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service