Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
1.04594
Calcon (C.I.15705)
metal indicator
Synonym(s):
Calcon (C.I.15705), 2-Hydroxy-1-(2-hydroxy-1-naphthylazo)naphthalene-4-sulfonic acid sodium salt
About This Item
Recommended Products
Product Name
Calcon (C.I.15705), metal indicator
Quality Level
form
solid
loss
≤10% loss on drying, 110°C
pH
9.4 (25 °C, 10 g/L in H2O)
solubility
20 g/L
bulk density
530 kg/m3
λmax
634-640 nm (buffer pH 12.2)
storage temp.
2-30°C
SMILES string
[Na+].[S](=O)(=O)([O-])C1=CC(=O)\C(=N\Nc3c4c(ccc3O)cccc4)\c2c1cccc2
InChI
1S/C20H14N2O5S.Na/c23-16-10-9-12-5-1-2-6-13(12)19(16)21-22-20-15-8-4-3-7-14(15)18(11-17(20)24)28(25,26)27;/h1-11,21,23H,(H,25,26,27);/q;+1/p-1/b22-20+;
InChI key
YJUGDMOGPJJWDG-QPNALZDCSA-M
Related Categories
Analysis Note
Absorption maximum λmax. (buffer pH 12.2): 634 - 640 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.015 g/l; buffer pH 12.2; calculated on dried substance): 250 - 400
Loss on drying (110 °C): ≤ 10 %
Suitability as indicator (for metal titration): passes test
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Helpful?
-
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdfHelpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service