Skip to Content
Merck
All Photos(1)

Key Documents

1.04506

Millipore

L-Hydroxyproline

for biochemistry

Synonym(s):

L-Hydroxyproline, L(-)-4-Hydroxypyrrolidine-2-carboxylic acid, Hyp

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO3
CAS Number:
Molecular Weight:
131.13
MDL number:
UNSPSC Code:
12352209
EC Index Number:
200-091-9
Pricing and availability is not currently available.

Quality Level

form

solid

pH

5.5-6.5 (20 °C, 50 g/L in H2O)

mp

274 °C

solubility

500 g/L

bulk density

610 kg/m3

SMILES string

N1C(CC(C1)O)C(=O)O

InChI

1S/C5H9NO3/c7-3-1-4(5(8)9)6-2-3/h3-4,6-7H,1-2H2,(H,8,9)

InChI key

PMMYEEVYMWASQN-UHFFFAOYSA-N

Analysis Note

Assay (perchloric acid titration, calculated on dried substance): ≥ 99.0 %
Identity (IR-spectrum): passes test
Appearance: white to almost white, crystalline powder or colorless crystals
Appearance of solution (100 g/l, water): clear and colorless
Spec. rotation (α 20/D, 50 g/l, hydrochloric acid 1 mol/l, calc. on dried substance): -47.5 to -46.0
Heavy metals (as Pb): ≤ 0.001 %
NH₄ (Ammonium): ≤ 0.01 %
Foreign amino acids: ≤ 0.3 %
Other ninhydrine positive substances(as Glycine): ≤ 0.1 %
Methanol (GC): ≤ 0.3 %
Loss on drying (105 °C; 3 h): ≤ 0.4 %

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service