1.04482
ICP Multi element standard solution X certified reference material
(23 elements in diluted nitric acid) for surface water testing, Certipur®
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
Quality Level
technique(s)
ICP: suitable
matrix
(in diluted nitric acid)
pH
1 (20 °C in water)
bp
100 °C/1013 hPa
density
1.007 g/cm3 at 20 °C
application(s)
environmental
food and beverages
personal care
(industry & testing)
(mining)
format
multi-component solution
storage temp.
15-25°C
General description
This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.
Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.
Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.
Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.
Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.
Packaging
100ml in HD-PE bottle
Analysis Note
Ca (calcium): 35000 µg/l
Mg (Magnesium): 15000 µg/l
Na (sodium): 8000 µg/l
K (potassium): 3000 µg/l
B (boron): 100 µg/l
Fe (iron): 100 µg/l
Mo (Molydenium): 100 µg/l
Sr (Strontium): 100 µg/l
As (arsenic): 50 µg/l
Ba (barium): 50 µg/l
Ni (Nickel): 50 µg/l
V (Vanadium): 50 µg/l
Zn (Zinc): 50 µg/l
Mn (Manganese): 30 µg/l
Co (cobalt): 25 µg/l
Pb (lead): 25 µg/l
Be (beryllium): 20 µg/l
Cd (cadmium): 20 µg/l
Cr (chromium): 20 µg/l
Cu (Copper): 20 µg/l
Bi (bismuth): 10 µg/l
Se (Selenium): 10 µg/l
Tl (Thallium): 10 µg/l
Mg (Magnesium): 15000 µg/l
Na (sodium): 8000 µg/l
K (potassium): 3000 µg/l
B (boron): 100 µg/l
Fe (iron): 100 µg/l
Mo (Molydenium): 100 µg/l
Sr (Strontium): 100 µg/l
As (arsenic): 50 µg/l
Ba (barium): 50 µg/l
Ni (Nickel): 50 µg/l
V (Vanadium): 50 µg/l
Zn (Zinc): 50 µg/l
Mn (Manganese): 30 µg/l
Co (cobalt): 25 µg/l
Pb (lead): 25 µg/l
Be (beryllium): 20 µg/l
Cd (cadmium): 20 µg/l
Cr (chromium): 20 µg/l
Cu (Copper): 20 µg/l
Bi (bismuth): 10 µg/l
Se (Selenium): 10 µg/l
Tl (Thallium): 10 µg/l
Other Notes
For a complete product listing of our Certipur® range of CRMs for ICP and AAS, technical information, and example certificates please visit our ICP & AAS standards website
Legal Information
Certipur is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service