Skip to Content
Merck

104132

SAFC

Cellvento® ModiFeed Prime COMP

greener alternative

chemically defined cell culture feed

Pharma Manufacturing

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213

Quality Level

description

shipped with cooling packs

product line

Cellvento®

form

dry powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

cell culture | mammalian: suitable (suspension)

greener alternative category

storage temp.

2-8°C

General description

Cellvento® ModiFeed Prime COMP is a highly concentrated, one-part, pH neutral feed added to replenish depleted nutrients required for cellular function and to extend the production phase in fed-batch mode. This single feed is highly concentrated, allowing for a reduction in volume of feed added to the medium thereby increasing product yield.The formulation has been specifically designed to provide increased process control and decreased complexity. Exclusion of glucose from the formulation allows for fed-batch process dictated regulation of this critical parameter. As a one-part feed, containing modified amino acids and amino acid derivatives, the complexity of hydration, storage, and feeding is drastically reduced.
We are committed to bringing you Greener Alternative Products, which belongs to one of the four categories of greener alternatives . This product is a Design for Sustainability (DfS) developed product. Click here to view its DfS scorecard.

Application

Cellvento® ModiFeed Prime COMP has been developed as a feed supplement to be used in combination with the production media, Cellvento® 4CHO COMP and EX-CELL® Advanced CHO, during fed-batch manufacturing processes. It has been designed to support optimal cell growth and high productivity of suspension CHO cell lines such as CHO-K1, CHO-DG44, CHO-S and CHO-GS, including cell lines derived using the CHOZN® platform Fed-batch (batch, perfusion)

Quantity

Formulated to contain 131.6 grams of powder per liter of medium.

Legal Information

CELLVENTO is a registered trademark of Merck KGaA, Darmstadt, Germany
CHOZN is a registered trademark of Merck KGaA, Darmstadt, Germany
EX-CELL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This case study walks through optimizing feed and media for upstream bioprocessing and how it translates to large-scale bioreactors.

Why optimize your upstream bioprocess media and feed? This article discusses the why and how of optimizing your media and feed to maximize cell viability and productivity.

Cell culture media are vital for bioprocessing therapeutics. This page describes the central role of our organization in authoring purpose-built GMP for cell culture media.

Cellvento® and EX-CELL® CHO cell culture media and feeds, optimized for monoclonal antibody & recombinant protein production in fed-batch & perfusion

Related Content

Learn more about cell culture media development for mAB development and manufacturing, and the products you need for this step.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service