Skip to Content
Merck
All Photos(3)

Key Documents

1.04008

Supelco

Formamide

EMPLURA®

Synonym(s):

Formic acid amide, Methane amide, Amide C1, Formic amide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCONH2
CAS Number:
Molecular Weight:
45.04
Beilstein:
505995
MDL number:
UNSPSC Code:
12352111
EC Index Number:
200-842-0
NACRES:
NA.21
Assay:
≥99% (N)
technique(s):
hybridization: suitable
bp:
210 °C (lit.)
vapor pressure:
0.08 mmHg ( 20 °C)
30 mmHg ( 129 °C)

vapor density

1.55 (vs air)

Quality Level

vapor pressure

0.08 mmHg ( 20 °C)
30 mmHg ( 129 °C)

product line

EMPLURA®

Assay

≥99% (N)

form

liquid

autoignition temp.

932 °F

potency

5325 mg/kg LD50, oral (Rat)
17000 mg/kg LD50, skin (Rabbit)

expl. lim.

2.7-19.0 % (v/v)

technique(s)

hybridization: suitable

impurities

≤0.3% Water (Karl Fischer)

refractive index

n20/D 1.447 (lit.)

pH

8-10 (20 °C, 200 g/L in H2O)

bp

210 °C (lit.)

mp

2-3 °C (lit.)

transition temp

flash point 175 °C

density

1.134 g/mL at 25 °C (lit.)

storage temp.

15-25°C

SMILES string

NC=O

InChI

1S/CH3NO/c2-1-3/h1H,(H2,2,3)

InChI key

ZHNUHDYFZUAESO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Formamide destabilizes nucleic acid duplexes and may be used, typically, at a concentration of 50%, in hybridization protocols requiring lower hybridization temperatures.

Analysis Note

Assay (N; Kjeldahl): ≥ 99.0 %
Identity (IR): passes test
Density (d 20 °C/ 4 °C): 1.132 - 1.135
Water (according to Karl Fischer): ≤ 0.3 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Repr. 1B - STOT RE 2 Oral

Target Organs

Blood

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

305.6 °F

Flash Point(C)

152 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service