Skip to Content
Merck

1.03668

SAFC

Parteck® M DPI

EMPROVE® EXPERT, Ph. Eur., BP, JP, USP, E 421

Pharma Manufacturing

Synonym(s):

D-Mannitol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14O6
CAS Number:
Molecular Weight:
182.17
MDL number:
UNSPSC Code:
12352107
E Number:
E421
EC Index Number:
200-711-8

Agency

BP
JP
Ph. Eur.
USP

Quality Level

product line

EMPROVE® EXPERT

form

fine powder

particle size

145-200 μm (d50)

pH

5-7 (20 °C, 100 g/L in H2O)

mp

164-169 °C

solubility

213 g/L

application(s)

liquid formulation
pharmaceutical
solid formulation

storage temp.

2-25°C

InChI

1S/C6H14O6/c7-1-3(9)5(11)6(12)4(10)2-8/h3-12H,1-2H2

InChI key

FBPFZTCFMRRESA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

With formulation challenges in mind, we have used particle engineering technologies to develop functional excipients specifically for solid dosage forms. The products in our Parteck® portfolio feature unique particle properties tailored for excellent performance in tableting processes, for specific drug delivery technologies, or for solubility enhancement.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Parteck® M DPI is a synthetic and versatile carrier option for your dry powder inhalation (DPI) formulation. Engineered from mannitol, Parteck® M DPI particles are designed to improve the flow and release characteristics of active pharmaceutical ingredients (APIs) in drugs delivered via dry, inhaled powders.
As Emprove® Expert product, Parteck® M DPI is specified with low low microbial and endotoxin limits, making is especially suitable for high-risk applications.

Features and Benefits

  • Particle distribution designed to improve flow and release characteristics in DPI application
  • Low water content & hygroscopicity
  • Synthesized from plant-based raw materials
  • 3-year shelf life
  • High API delivery to the lungs

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
PARTECK is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service