Skip to Content
Merck
All Photos(1)

Key Documents

1.02768

Supelco

Copper(II) oxide

granular for analysis EMSURE®

Synonym(s):

Copper(II) oxide, Copper monoxide granulated

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CuO
CAS Number:
Molecular Weight:
79.55
MDL number:
UNSPSC Code:
12352303
EC Index Number:
215-269-1
Assay:
≥99.0% (complexometric)
form:
solid
Pricing and availability is not currently available.

Quality Level

product line

EMSURE®

Assay

≥99.0% (complexometric)

form

solid

potency

470 mg/kg LD50, oral (Rat)

impurities

≤0.02% Substances insoluble in nitric acid

pH

7 (20 °C, 50 g/L in H2O, slurry)

mp

1326 °C (decomposition)

density

6.48 g/cm3 at 25 °C

bulk density

500 kg/m3

anion traces

chloride (Cl-): ≤0.005%
sulfur compounds (as SO42-): ≤0.015%

cation traces

Cr: ≤0.05%
Fe: ≤0.05%
N: ≤0.002%
Pb: ≤0.05%
excess alkali:, passes test

storage temp.

no temp limit

InChI

1S/Cu.O

InChI key

QPLDLSVMHZLSFG-UHFFFAOYSA-N

Analysis Note

Assay (complexometric): ≥ 99.0 %
Substances insoluble in nitric acid: ≤ 0.02 %
Free Alkali: passes test
Chloride (Cl): ≤ 0.005 %
Total sulfur (as sulphates): ≤ 0.015 %
Total nitrogen (N): ≤ 0.002 %
Cr (Chromium): ≤ 0.05 %
Fe (Iron): ≤ 0.05 %
Pb (Lead): ≤ 0.05 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service