Skip to Content
Merck
All Photos(1)

Documents

1.02149

Millipore

Contact TVC

Envirocheck, Contact total viable count coated with Nutrient Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.74

material

plastic cap
plastic slide
plastic tube

Quality Level

product line

Envirocheck

manufacturer/tradename

Envirocheck

technique(s)

cell counting: suitable (total viable count)

application(s)

food and beverages
surface monitoring
water monitoring

storage temp.

2-30°C

suitability

bacteria (total viable count)

General description

(Total Viable Counts) Envirocheck
Envirocheck contact total viable count, a semi-quantitative detection of microorganisms on surfaces and in liquids, are flexible paddles that are coated with Nutrient Agar to determine the total viable count of bacteria in sample. Slide A is Coated with Nutrient Agar containing 0.05% Triphenyl Tetrazolium Chloride (TTC) and Slide B Coated with Nutrient Agar. The growth of bacteria on slide A is indicated by red colonies due to the formation of red formazan dye when TTC is reduced.

Application

Envirocheck contact slides can be used for both surface and liquid testing in industrial and environmental monitoring.

Analysis Note

Count the number of colonies on both sides of the paddle- results should not differ by a factor greater than 2, where the colonies on side A will appear mainly in red. In the presence of TTC a few microorganisms may grow less profusely. Always go with the higher counts in case they are not identical on both sides. This represents the microbial load for the test.
Sterility: passes test
pH-value/side one (25 °C): 6.7 - 7.3
pH-value/side two (25 °C): 6.7 - 7.3
Growth/side one
- Escherichia coli ATCC 8739 (WDCM 00012):
good
- Staphylococcus aureus ATCC 6538 (WDCM 00032): fair/good
- Candida albicans ATCC 10231 (WDCM 00054): none/poor
- Bacillus subtilis ATCC 6633 (WDCM 00003): good
- Pseudomonas aeruginosa ATCC 9027 (WDCM 00026): good
- Aspergillus brasiliensis (formerly A. niger) ATCC 16404 (WDCM 00053): good
Growth/side two
- Escherichia coli ATCC 8739 (WDCM 00012):
good
- Staphylococcus aureus ATCC 6538 (WDCM 00032): good
- Candida albicans ATCC 10231 (WDCM 00054): good
- Bacillus subtilis ATCC 6633 (WDCM 00003): good
- Pseudomonas aeruginosa ATCC 9027 (WDCM 00026): good
- Aspergillus brasiliensis (formerly A. niger) ATCC 16404 (WDCM 00053): good
Incubation: 48 hrs.; 35 °C; aerobic; Aspergillus up to 72 hrs.

Legal Information

ENVIROCHECK is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Related Content

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service