Skip to Content
Merck

1.02110

SAFC

Calcium hydroxide

EMPROVE® ESSENTIAL, Ph. Eur., BP, JP, USP, FCC, E 526

Pharma Manufacturing

Synonym(s):

Calcium hydroxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ca(OH)2
CAS Number:
Molecular Weight:
74.09
MDL number:
UNSPSC Code:
12352305
E Number:
E526
EC Index Number:
215-137-3

Agency

BP
JP
Ph. Eur.
USP

Quality Level

reg. compliance

FCC

product line

EMPROVE® ESSENTIAL

form

solid

pH

12.6 (20 °C in H2O, saturated solution)

mp

550 °C (decomposition)

density

2.24 g/cm3 at 20 °C

application(s)

liquid formulation
pharma/biopharma processes
pharmaceutical
semi-solid formulation
solid formulation

storage temp.

2-30°C

SMILES string

[Ca+2].[O-H].[O-H]

InChI

1S/Ca.2H2O/h;2*1H2/q+2;;/p-2

InChI key

AXCZMVOFGPJBDE-UHFFFAOYSA-L

Related Categories

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Calcium hydroxide is used as pH adjuster in pharmaceutical formulations and cosmetics. It is also used as additive for food and beverage.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service