Skip to Content
Merck
All Photos(2)

Key Documents

1.01830

Supelco

Acetic acid (glacial)

100%, for analysis EMPARTA® ACS

Synonym(s):

Acetic acid, Ethanoic acid, Glacial acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2H
CAS Number:
Molecular Weight:
60.05
Beilstein:
506007
MDL number:
UNSPSC Code:
12352106
EC Index Number:
200-580-7
NACRES:
NA.21

product name

Acetic acid (glacial) 100%, for analysis EMPARTA® ACS

grade

ACS reagent

Quality Level

vapor density

2.07 (vs air)

vapor pressure

15.4 hPa ( 20 °C)

product line

EMPARTA®

Assay

≥99.7% (acidimetric)

form

liquid

autoignition temp.

485 °C
800 °F

potency

3310 mg/kg LD50, oral (Rat)

expl. lim.

16 %, 92 °F
4 %, 59 °F

impurities

≤0.0004 meq/g Titratable base
≤100 ppm Acetic anhydride

evapn. residue

≤10 ppm

color

APHA: ≤10

refractive index

n20/D 1.371 (lit.)

pH

2.5 (20 °C, 50 g/L in H2O)

kinematic viscosity

1.17 cSt(20 °C)

bp

117-118 °C (lit.)

mp

16.2 °C (lit.)

transition temp

flash point 39 °C

solubility

soluble 602.9 g/L

density

1.04 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤1 ppm
sulfate (SO42-): ≤1 ppm

cation traces

Fe: ≤0.2 ppm
heavy metals (as Pb): ≤0.5 ppm

storage temp.

15-25°C

SMILES string

[F2C(F2C)13F3C]C(O)=O

InChI

1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)

InChI key

QTBSBXVTEAMEQO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Screening Ultra-Stable (Phenazine)dioxyalkanocic Acids with Varied Water-Solubilizing Chain Lengths for High-Capacity Aqueous Redox Flow Batteries: This study explores the application of phenazine-based compounds, where acetic acid′s derivatives act as soluble mediators in redox flow batteries, offering insights into optimizing battery electrolytes for enhanced capacity and stability (Liu et al., 2024).

  • Ambient Aqueous Synthesis of Imine-Linked Covalent Organic Frameworks (COFs) and Fabrication of Freestanding Cellulose Nanofiber@COF Nanopapers: This research demonstrates the use of acetic acid in the eco-friendly synthesis of imine-linked covalent organic frameworks, highlighting its role in promoting green chemistry practices within material science (Kong et al., 2024).

  • Green, Safe, and Reliable Synthesis of Bimetallic MOF-808 Nanozymes With Enhanced Aqueous Stability and Reactivity for Biological Applications: Glacial acetic acid is used to modulate the pH during the synthesis of MOF-808 nanozymes, improving their biological compatibility and stability for potential therapeutic applications (Simms et al., 2024).

  • Dual-Functional Manganese-Doped ZnO-MOF Hybrid Material with Enhanced Hydrolytic Stability: In this study, acetic acid is integral in the synthesis of a ZnO-based metal-organic framework, enhancing its hydrolytic stability and functionality as a sensor for environmental monitoring (Asadevi et al., 2023).

  • Extraction of Nucleotides from Dietary Supplements by Newly Synthesized Adsorbents: The research utilizes acetic acid in the extraction process of nucleotides from dietary supplements, showcasing its efficacy in increasing the yield and purity of extracted compounds (Studzińska et al., 2023).

Analysis Note

Assay (acidimetric): ≥ 99.7 %
Color: ≤ 10 Hazen
Titratable base: ≤ 0.0004 meq/g
Acetic anhydride: ≤ 100 ppm
Chloride (Cl): ≤ 1 ppm
Heavy metals (as Pb): ≤ 0.5 ppm
Sulfate (SO₄): ≤ 1 ppm
Fe (Iron): ≤ 0.2 ppm
Substances reducing potassium dichromate: passes test
Substances reducing potassium permanganate: passes test
Evaporation residue: ≤ 10 ppm
Dilution test: passes test

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

102.2 °F

Flash Point(C)

39 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service