Skip to Content
Merck
All Photos(1)

Key Documents

1.01167

Sigma-Aldrich

Amido black 10 B (C.I.20470)

for electrophoresis

Synonym(s):

Amido black 10 B (C.I.20470), Black acid 1, Amido black 10 B (C.I. 20470), Naphthol blue black

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H14N6Na2O9S2
CAS Number:
Molecular Weight:
616.49
MDL number:
UNSPSC Code:
41105305
EC Index Number:
213-903-1
Pricing and availability is not currently available.

Quality Level

form

solid

potency

>5000 mg/kg LD50, oral (Rat)

composition

Dye, ≥80% spectrophotometric assay

pH

9.0 (25 °C, 10 g/L in H2O)

solubility

10 g/L

bulk density

670 kg/m3

εmax

750-960 at 617-620 nm in water at 0.01 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

InChI

1S/C22H16N6O9S2.2Na/c23-19-18-12(11-17(39(35,36)37)21(22(18)29)27-24-13-4-2-1-3-5-13)10-16(38(32,33)34)20(19)26-25-14-6-8-15(9-7-14)28(30)31;;/h1-11,24H,23H2,(H,32,33,34)(H,35,36,37);;/q;2*+1/p-2/b26-25?,27-21+;;

InChI key

HKBVRFLHNUEVRO-DWTBGCDMSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Amido black 10 B (C.I. 20470) for electrophoresis, is used for staining connective tissue, in electrophoresis, and as a redox indicator. Proteins possess free amino and carboxyl groups and are charged positively or negatively in aqueous solutions, depending on the pH. In an alkaline milieu they are negatively charged and migrate in the electric field towards the anode at a speed that depends on the number of charges they carry. This difference in migration velocities makes it possible to separate the serum protein mixture into groups of individual components.The separated protein fractions are stained with amido black and measured photometrically or densitometrically. As a rule, five fractions are observed after staining - albumin, alpha 1- and alpha 2-globulins, beta 1- and beta 2-globulins, and beta-globulins - where albumin has the highest migration velocity and gamma-globulin the lowest. Differentiation of serum proteins by electrophoresis is important for the diagnosis and prognosis of numerous diseases. Changes in protein composition can be found e. g. in inflammatory diseases, tumors, liver diseases, antibody deficiency syndromes or disturbances in protein balance. As an IVD product and CE registered, it can be used hassle-free in diagnostics and laboratory accreditations. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Analysis Note

Dye content (spectrophotometrically): ≥ 80 %
Identity
- sodium:
passes test
- UV/VIS-Spectrum: passes test
Absorption maximum λ max (water): 617 - 620 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.01 g/l; water): 750 - 960
TLC-Test: passes test
Loss on drying (110 °C): ≤ 10 %
Suitability for electrophoresis: passes test
Suitability as redox indicator: passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service