Skip to Content
Merck
All Photos(1)

Key Documents

1.01086

Supelco

Aluminium nitrate nonahydrate

EMPLURA®

Synonym(s):

Aluminium nitrate nonahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al(NO3)3 · 9 H2O
CAS Number:
Molecular Weight:
375.13
MDL number:
UNSPSC Code:
12352302
EC Index Number:
236-751-8
NACRES:
NA.21
form:
solid
solubility:
41.9 g/L
Pricing and availability is not currently available.

Quality Level

product line

EMPLURA®

Assay

≥95.0% (complexometric)

form

solid

potency

3671 mg/kg LD50, oral (Rat)

impurities

≤0.5% Substances not precipitated by ammonia (as sulfate)

pH

2.0-4.0 (20 °C, 50 g/L in H2O)

mp

73 °C

solubility

41.9 g/L

density

1.72 g/cm3 at 20 °C

bulk density

880 kg/m3

anion traces

chloride (Cl-): ≤0.005%
sulfate (SO42-): ≤0.03%

cation traces

Fe: ≤0.01%
heavy metals (as Pb): ≤0.005%

storage temp.

2-30°C

SMILES string

[Al]1(O[N+](=O)O1)O[N+](=O)[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O

InChI

1S/Al.3NO3.9H2O/c;3*2-1(3)4;;;;;;;;;/h;;;;9*1H2/q+3;3*-1;;;;;;;;;

InChI key

SWCIQHXIXUMHKA-UHFFFAOYSA-N

Related Categories

Application


  • Surface Acidity and Hydrophilicity of Coprecipitated Al(2)O(3)-SiO(2) Xerogels Prepared from Aluminium Nitrate Nonahydrate and Tetraethylorthosilicate.: This study explores the properties of xerogels, highlighting their relevance in catalysis and materials science, emphasizing the role of aluminium nitrate nonahydrate in modifying surface characteristics (Okada et al., 1999).

  • How do layered double hydroxides evolve. First in situ insights into their synthesis processes.: Discusses the synthesis of layered double hydroxides using aluminium nitrate nonahydrate, providing valuable insights for chemists involved in materials chemistry and nanotechnology (Terraschke et al., 2022).

  • Vibrational models for a crystal with 36 water molecules in the unit cell: IR spectra from experiment and calculation.: Presents vibrational analysis of crystal structures involving aluminium nitrate nonahydrate, which can aid in understanding molecular interactions and crystal engineering (Hermansson et al., 2015).

  • Antioxidant activity and cytotoxicity of Jerusalem artichoke tubers and leaves extract on HaCaT and BJ fibroblast cells.: While focused on plant extracts, this research uses methodologies and analysis techniques that can be insightful for chemists working with complex organic matrices and their interactions with inorganic ions like those from aluminium nitrate nonahydrate (Zagórska-Dziok et al., 2018).

  • Comparative aluminium mobilizing actions of several chelators in aluminium-loaded uraemic rats.: This study provides data on the bioactivity of chelators, including those that might interact with aluminium nitrate nonahydrate, relevant for medical and pharmaceutical chemists focusing on metal toxicity and treatment strategies (Corbella et al., 1994).

Analysis Note

Assay (complexometric): ≥ 95.0 %
Chloride (Cl): ≤ 0.005 %
Sulfate (SO₄): ≤ 0.03 %
Heavy metals (as Pb): ≤ 0.005 %
Fe (Iron): ≤ 0.01 %
Substances not precipitated by ammonia (as sulfate): ≤ 0.5 %

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service