Skip to Content
Merck
All Photos(3)

Documents

07-582-I

Sigma-Aldrich

Anti-phospho-IRF-3 (Ser396) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Interferon regulatory factor 3, IRF-3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

porcine (based on 100% sequence homology), rhesus macaque (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer396)

Gene Information

human ... IRF3(3661)
pig ... Irf3(396656)
rhesus monkey ... Irf3(718970)

General description

Interferon regulatory factors (IRFs) comprise a family of transcription factors that function with the Jak/Stat pathway to regulate interferon (IFN) and IFN-inducible gene expression in response to viral infection. IRF-3 can inhibit cell growth and plays a critical role in controlling the expression of genes in the innate immune response. In unstimulated cells, IRF-3 is present in the cytoplasm. Viral infection results in phosphorylation of IRF-3 and leads to its translocation to the nucleus where it activates promoters containing IRF-3-binding sites. Phosphorylation of IRF-3 occurs at a cluster of C-terminal serine and threonine residues (between 385 and 405) leading to its association with the p300/CBP coactivator protein that promotes DNA binding and transcriptional activity.

Specificity

This antibody recognizes IRF-3 phosphorylated at Ser396.

Immunogen

Epitope: SNS → ANA: Mutation of this residue results in loss of viral-induced phosphorylation of IRF-3.
KLH-conjugated linear peptide corresponding to human IRF-3 phosphorylated at Ser396.

Application

Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
Use Anti-phospho-IRF-3 (Ser396) Antibody (Rabbit Polyclonal Antibody) validated in WB to detect phospho-IRF-3 (Ser396) also known as Interferon regulatory factor 3.

Quality

Evaluated by Western Blot in untreated and sendai virus treated A549 cell lysates.

Western Blot Analysis: A 1:1,000 dilution of this antibody detected phospho-IRF-3 (Ser396) in 10 µg of sendai virus treated A549 cell lysate.

Target description

~50 kDa observed. Uncharacterized bands may appear at ~40 kDa and ~80 kDa in some lysates.

Linkage

Replaces: 07-582

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Untreated and sendai virus treated A549 cell lysates

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mitsuaki Mori et al.
The Journal of biological chemistry, 279(11), 9698-9702 (2004-01-03)
Interferon regulatory factor (IRF)-3 is a critical transcription factor regulating innate immune responses against viral and bacterial infections. Signals activated by various pathogens are integrated by IRF-3 kinase, resulting in the specific phosphorylation of IRF-3 in the cytoplasm. This phosphorylation
Cortisol is a potent modulator of lipopolysaccharide-induced interferon signaling in macrophages.
Anja M Billing,Fred Fack,Jonathan D Turner,Claude P Muller
Innate Immunity null
Xiaokun Liu et al.
Frontiers in immunology, 8, 350-350 (2017-04-11)
Influenza virus NS2 is well known for its role in viral ribonucleoprotein nuclear export; however, its function has not been fully understood. A recent study showed that NS2 might interact with HIST1H1C (H1C, H1.2). Histones have been found to affect

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service