Skip to Content
Merck
All Photos(1)

Key Documents

07-539

Sigma-Aldrich

Anti-acetyl-Histone H3 (Lys 4) Antibody

Upstate®, from rabbit

Synonym(s):

H3K4Ac, Histone H3 (acetyl K4)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

ChIP: suitable (ChIP-seq)
dot blot: suitable
inhibition assay: suitable (peptide)
multiplexing: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

acetylation (Lys 4)

Gene Information

human ... H3F3B(3021)

Specificity

Broad species cross reactivity is expected
Histone H3 when acetylated on Lys4

Immunogen

KLH-conjugated, synthetic peptide containing the sequence ...RT[AcK]Q... in which [AcK] corresponds to acetyl lysine 4 of human histone H3

Application

Suggested dilutions:
Western Blot: 1:1000 - 1:10,000
Anti-acetyl-Histone H3 (Lys 4) Antibody is a rabbit polyclonal antibody for detection of acetyl-Histone H3 (Lys 4) also known as H3K4Ac, Histone H3 (acetyl K4) & has been validated in Mplex, WB, PIA, ChIP, ChIP-seq, DB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

Routinely evaluated by immunoblot.

Target description

Mr ~17kDa

Physical form

100μl of immunoaffinity purified rabbit IgG in a buffer with 0.1M Tris-glycine, pH 7.4 and 0.15M NaCl, and containing 0.05% sodium azide and 30% glycerol
ImmunoAffinity Purified

Storage and Stability

1 year at -20°C from date of shipment

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Federico Forneris et al.
The Journal of biological chemistry, 281(46), 35289-35295 (2006-09-22)
Human lysine-specific demethylase (LSD1) is a chromatin-modifying enzyme that specifically removes methyl groups from mono- and dimethylated Lys4 of histone H3 (H3-K4). We used a combination of in vivo and in vitro experiments to characterize the substrate specificity and recognition
Oral administration of the pimelic diphenylamide HDAC inhibitor HDACi 4b is unsuitable for chronic inhibition of HDAC activity in the CNS in vivo.
Beconi, M; Aziz, O; Matthews, K; Moumne, L; O'Connell, C; Yates, D; Clifton, S; Pett et al.
Testing null
Flore Beurton et al.
Nucleic acids research, 47(21), 11164-11180 (2019-10-12)
The CFP1 CXXC zinc finger protein targets the SET1/COMPASS complex to non-methylated CpG rich promoters to implement tri-methylation of histone H3 Lys4 (H3K4me3). Although H3K4me3 is widely associated with gene expression, the effects of CFP1 loss vary, suggesting additional chromatin
Ya-Wei Chen et al.
Cancer, 119(24), 4259-4267 (2013-12-05)
Patterns of global histone modifications have been suggested to be predictors of clinical outcome in many cancers. However, the role of global histone modification patterns in oral squamous cell carcinoma (OSCC) is unclear. A retrospective clinicopathologic analysis was undertaken of
Runsheng He et al.
Nucleic acids research, 45(11), 6427-6441 (2017-04-13)
Epigenetic regulation of chromatin plays a critical role in controlling embryonic stem (ES) cell self-renewal and pluripotency. However, the roles of histone demethylases and activating histone modifications such as trimethylated histone 3 lysine 4 (H3K4me3) in transcriptional events such as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service