Skip to Content
Merck
All Photos(1)

Key Documents

07-503

Sigma-Aldrich

Anti-p40-phox Antibody

Upstate®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... NCF4(4689)
rat ... Ncf4(500904)

Specificity

p40-phox

Immunogen

C-terminal 6HIS-tagged fusion protein corresponding to full length human p40-phox

Application

Anti-p40-phox Antibody is an antibody against p40-phox for use in WB.
Research Category
Neuroscience
Research Sub Category
Oxidative Stress

Quality

routinely evaluated by immunoblot in RIPA lysates of HL-60 cells.

Target description

40kDa

Physical form

Format: Purified
Protein A purified
Protein A purified antibody in 0.1M Tris-Glycine, 0.15M NaCl, 0.05% Sodium Azide, pH 7.4.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna-Lena Neehus et al.
Journal of clinical immunology, 42(6), 1244-1253 (2022-05-19)
Autosomal recessive (AR) PKCδ deficiency is a rare inborn error of immunity (IEI) characterized by autoimmunity and susceptibility to bacterial, fungal, and viral infections. PKCδ is involved in the intracellular production of reactive oxidative species (ROS). We studied a 5-year
A fluorescently tagged C-terminal fragment of p47phox detects NADPH oxidase dynamics during phagocytosis.
Li, XJ; Tian, W; Stull, ND; Grinstein, S; Atkinson, S; Dinauer, MC
Molecular Biology of the Cell null
Regulation of renal epithelial tight junctions by the von Hippel-Lindau tumor suppressor gene involves occludin and claudin 1 and is independent of E-cadherin.
Harten SK, Shukla D, Barod R, Hergovich A, Balda MS, Matter K, Esteban MA, Maxwell PH.
Molecular Biology of the Cell null
F B Wientjes et al.
The Biochemical journal, 296 ( Pt 3), 557-561 (1993-12-15)
The NADPH oxidase generates superoxide in phagocytic cells. It is important for immunity and its deficiency leads to chronic granulomatous disease (CGD). It consists of a membrane-bound flavocytochrome b that lies dormant until activated by the translocation to the plasma
Hassan Dakik et al.
Antioxidants (Basel, Switzerland), 10(3) (2021-04-04)
In acute myeloid leukemia (AML), a low level of reactive oxygen species (ROS) is associated with leukemic stem cell (LSC) quiescence, whereas a high level promotes blast proliferation. ROS homeostasis relies on a tightly-regulated balance between the antioxidant and oxidant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service