Skip to Content
Merck
All Photos(1)

Key Documents

07-415

Sigma-Aldrich

Anti-Sam 68 Antibody

serum, Upstate®

Synonym(s):

Anti-Sam68, Anti-p68

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human, rat

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KHDRBS1(10657)

Specificity

Sam 68

Immunogen

Peptide corresponding to residues 331-348 of human Sam 68

Application

Anti-Sam 68 Antibody is a high quality Rabbit Polyclonal Antibody for the detection of Sam 68 & has been validated in IP, WB, ICC.
Research Category
Signaling
Research Sub Category
Insulin/Energy Signaling

Quality

routinely evaluated by immunoblot in RIPA lysates from HeLa cells

Target description

68kDa

Physical form

0.1M Trisglycine,pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Antiserum

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jawed Iqbal et al.
PLoS pathogens, 12(10), e1005967-e1005967 (2016-10-21)
IFI16 (gamma-interferon-inducible protein 16), a predominantly nuclear protein involved in transcriptional regulation, also functions as an innate immune response DNA sensor and induces the IL-1β and antiviral type-1 interferon-β (IFN-β) cytokines. We have shown that IFI16, in association with BRCA1
Xun Hui Yeo et al.
Communications biology, 6(1), 660-660 (2023-06-23)
AXL is a receptor tyrosine kinase that is often overexpressed in cancers. It contributes to pathophysiology in cancer progression and therapeutic resistance, making it an emerging therapeutic target. The first-in-class AXL inhibitor bemcentinib (R428/BGB324) has been granted fast track designation
Oyindamola Awe et al.
Pregnancy hypertension, 22, 196-203 (2020-10-18)
The goal of this study was to determine if an axis of placental gene expression associated with early onset and severe preeclampsia (EOSPE) was operative in term pregnancy and correlated with vitamin D sufficiency. qPCR analysis of NKX2-5, SAM68, sFLT1
Clément Chapat et al.
Nucleic acids research, 45(14), 8508-8523 (2017-06-08)
The CCR4-associated factor CAF1, also called CNOT7, is a catalytic subunit of the CCR4-NOT complex, which has been implicated in all aspects of the mRNA life cycle, from mRNA synthesis in the nucleus to degradation in the cytoplasm. In human
Yannick D Benoit et al.
Cell chemical biology, 24(7), 833-844 (2017-06-27)
Targeting of human cancer stem cells (CSCs) requires the identification of vulnerabilities unique to CSCs versus healthy resident stem cells (SCs). Unfortunately, dysregulated pathways that support transformed CSCs, such as Wnt/β-catenin signaling, are also critical regulators of healthy SCs. Using

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service