Skip to Content
Merck
All Photos(1)

Key Documents

07-1801

Sigma-Aldrich

Anti-G3BP1 Antibody

serum, from rabbit

Synonym(s):

ATP-dependent DNA helicase VIII, GAP SH3 domain-binding protein 1, GAP binding protein, GTPase activating protein (SH3 domain) binding protein 1, Ras-GTPase-activating protein SH3-domain-binding protein, RasGAP-associated endoribonuclease G3BP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

species reactivity (predicted by homology)

rat (100% immunogen homology)

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... G3BP1(10146)
mouse ... G3Bp1(27041)

General description

G3BP1 (Ras-GTPase activating protein SH3-domain binding protein 1) directly associates with SH3 domains of GTPase activating proteins (GAPs). It was first discovered by its binding to RasGAP. G3BPs have been shown to be involved in a number of mitogenic signaling pathways. G3BP1 is also known to be over expressed in many human cancers (Kim, 2007) and its expression appears to be inversely correlated with PTEN expression (Kim, 2007). Interestingly, G3BP1, along with G3BP2, has been shown to bind to the C-terminus of p53, playing a crucial role in its activity (Kim, 2007). Furthermore, the G3BPs have two traditional RNA binding motifs.

Specificity

Not tested on other species.
This antibody detects G3BP1 at its C-terminal region.

Immunogen

Epitope: C-terminus
Synthetic peptide corresponding to C-terminal region of human G3BP1.

Application

Anti-G3BP1 Antibody detects level of G3BP1 & has been published & validated for use in WB & IP.
Research Category
Signaling
Research Sub Category
RNA Binding Protein (RBP)

Quality

Evaluated by western blot on HeLa cell lysates.

Western Blot Analysis:
1:1,000 dilution of this antibody was used to detect G3BP1 in HeLa, 293T and S10 cell lysates.

Target description

68 kDa

Physical form

Rabbit polyclonal serum with 0.05% NaN3.

Storage and Stability

Stable for 1 year at -20ºC from date of receipt.

Analysis Note

Control
HeLa cell lysates.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Oi et al.
Oncogene, 34(20), 2660-2671 (2014-07-08)
Resveratrol (trans-3,5,4'-truhydroxystilbene) possesses a strong anticancer activity exhibited as the induction of apoptosis through p53 activation. However, the molecular mechanism and direct target(s) of resveratrol-induced p53 activation remain elusive. Here, the Ras-GTPase-activating protein SH3 domain-binding protein 1 (G3BP1) was identified
Soumitra Ghosh et al.
EBioMedicine, 2(11), 1785-1798 (2016-02-13)
Microglial cells in the brains of Alzheimer's patients are known to be recruited to amyloid-beta (Aβ) plaques where they exhibit an activated phenotype, but are defective for plaque removal by phagocytosis. In this study, we show that microglia stressed by
Kalle Kipper et al.
Journal of molecular biology, 434(20), 167801-167801 (2022-08-30)
The polarized cell morphology of neurons dictates many neuronal processes, including the axodendridic transport of specific mRNAs and subsequent translation. mRNAs together with ribosomes and RNA-binding proteins form RNA granules that are targeted to axodendrites for localized translation in neurons.
Chelsea J Webber et al.
Human molecular genetics, 32(20), 2966-2980 (2023-07-31)
Aggregation of TAR DNA-binding protein 43 kDa (TDP-43) is thought to drive the pathophysiology of amyotrophic lateral sclerosis and some frontotemporal dementias. TDP-43 is normally a nuclear protein that in neurons translocates to the cytoplasm and can form insoluble aggregates upon
Shangmei Hou et al.
Journal of virology, 91(16) (2017-06-09)
Zika virus (ZIKV), a member of the Flaviviridae family, has recently emerged as an important human pathogen with increasing economic and health impact worldwide. Because of its teratogenic nature and association with the serious neurological condition Guillain-Barré syndrome, a tremendous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service