Skip to Content
Merck
All Photos(1)

Key Documents

07-1407

Sigma-Aldrich

Anti-GLUT-8 Antibody, CT

from rabbit

Synonym(s):

Glucose transporter type 8, Glucose transporter type X1, glucose transporter X1, solute carrier family 2 (facilitated glucose transporter)

member 8, solute carrier family 2 (facilitated glucose transporter),

member 8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

species reactivity (predicted by homology)

mouse

technique(s)

immunohistochemistry: suitable (paraffin)

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SLC2A8(29988)
mouse ... Slc2A8(56017)

Related Categories

General description

GLUT-8 is a glucose transporter facilitated by the regulation of insulin, binding cytochalasin B such that the orientation is glucose-inhibitive. In appearance, the transporter seems to be a dual-specific sugar transporter since fructose can also be inhibitive. GLUT-8 is expressed largely in the testis, where down-regulation is facilitated by estrogen. However, GLUT-8 is not highly expressed in testicular carcinoma. Lower levels of GLUT-8 are present in most other tissues.

Specificity

Recognizes GLUT-8.

Immunogen

An 11 amino acid sequence near the C-terminus of mouse Glut-8 (Doege et al. 2000; Carayannopoulos et al. 2000)
Epitope: C-terminus

Application

Anti-GLUT-8 Antibody, C-terminus detects level of GLUT-8 & has been published & validated for use in IH(P).
Research Category
Signaling
Research Sub Category
Insulin/Energy Signaling

Quality

Evaluated by Immunohistochemistry in human kidney tissue
Immunohistochemistry Analysis: 1:100 dilution of this lot detected GLUT-8 in human kidney tissue.

Target description

Approx. 50 kDa

Linkage

Replaces: AB3140

Physical form

Format: Purified
ImmunoAffinity Purified
Liquid in PBS with 0.1% BSA.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 1 year from date of receipt. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
Human kidney tissue.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rachel E Bennett et al.
Acta neuropathologica communications, 8(1), 137-137 (2020-08-20)
Microangiopathy, including proliferation of small diameter capillaries, increasing vessel tortuosity, and increased capillary blockage by leukocytes, was previously observed in the aged rTg4510 mouse model. Similar gene expression changes related to angiogenesis were observed in both rTg4510 and Alzheimer's disease
Jimena Laporta et al.
PloS one, 8(2), e57847-e57847 (2013-03-08)
The aim of this experiment was to demonstrate the ability of feeding serotonin (5-HT; 5-hydroxytryptamine) precursors to increase 5-HT production during the transition from pregnancy to lactation and the effects this has on maternal energy metabolism in the liver and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service