Skip to Content
Merck
All Photos(1)

Key Documents

07-1318

Sigma-Aldrich

Anti-phospho-JAK2 (Tyr317) Antibody

serum, from rabbit

Synonym(s):

Janus kinase 2, Janus kinase 2 (a protein tyrosine kinase), tyrosine-protein kinase JAK2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pTyr317)

Gene Information

human ... JAK2(3717)

General description

JAK2 (Janus Kinase 2) belongs to the emerging family of non-receptor Janus tyrosine kinases, which regulate a spectrum of cellular functions downstream of activated cytokine receptors in the lympho-hematopoietic system. Immunological stimuli, such as interferons and cytokines, induce recruitment of Stat transcription factors to cytokine receptor-associated JAK2. JAK2 then phosphorylates proximal Stat factors, which subsequently dimerize, translocate to the nucleus and bind to CIS elements upstream of target gene promoters to regulate transcription. The canonical JAK/Stat pathway is integral to maintaining a normal immune system by stimulating proliferation, differentiation, survival and host resistance to pathogens. Altering JAK/Stat signaling to reduce cytokine induced pro-inflammatory responses represents an attractive target for anti-inflammatory therapies.

Specificity

This antibody recognizes phosphorylated JAK2.

Immunogen

Epitope: Tyr317
KLH-conjugated recombinant protein selected from residues corresponding to Tyr317 of JAK2.

Application

Detect phospho-JAK2 (Tyr317) using this Anti-phospho-JAK2 (Tyr317) Antibody validated for use in WB.
Research Category
Signaling

Quality

Evaluated by Western Blot in JAK2 IP lysate (see below).
Western Blot Analysis: A 1:500 dilution of this antibody detected phospho-JAK2 (Tyr317) on JAK2 IP lysate.

Target description

130 kDa

Physical form

Rabbit serum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chengliang Yuan et al.
Journal of biochemical and molecular toxicology, 36(1), e22943-e22943 (2021-11-02)
Eriocitrin, a lemons flavanone, exhibits several biological properties, antiproliferative, and proapoptotic effects. However, its molecular mechanical action is not entirely clarified. Oxidative stress causes abnormal stimulation of signal transducer and activator of transcription 3 (STAT3) and c-Jun NH2-terminal kinase (JNK), p38

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service