Skip to Content
Merck
All Photos(2)

Key Documents

06-821-I

Sigma-Aldrich

Anti-phospho-PKCepsilon Antibody (Ser729)

from rabbit, purified by affinity chromatography

Synonym(s):

PKCepsilon, PKCe, Protein kinase C epsilon type, nPKC-epsilon

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human, mouse

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer729)

Gene Information

human ... PRKCE(5581)

General description

Protein Kinase C epsilon (PKC epsilon) is an ~87 kDa (migrates predominantly as 95 kDa in quiescient cells) member of the novel group (nPKCs: sensitive to diacylglycerol, phosphatidylserine, and phorbol esters) of the PKC family of serine/threonine kinases that are involved in a wide range of physiological processes including mitogenesis, cell survival, metastasis and transcriptional regulation. PKC epsilon is an actin cytoskeleton regulating protein whose overexpression confers an oncogenic phenotype by affecting the ras signaling cascade. PKC epsilon is phosphorylated by PDK1 on threonine 566 and is then autophosphorylated on the turn motif threonine 710 and the hydrophobic motif serine 729. Catalytically inactive PKC epsilon is not phosphorylated on serine 729. Phosphorylation of serine 729 is mediated by mTOR-dependent pathways, plays an important role in regulating PKC epsilon subcellular localization, and is responsible for the appearance of the 92 kDa PKC epsilon species.

Specificity

This antibody demonstrates cross-reactivity against the PKCbeta (Ser661) isoform, PKCdelta (Ser655) isoform, and possibly PKCalpha (Ser729), all of which contain the GF(pS)Y motif.

Immunogen

Epitope: Phosphorylated Ser729
KLH-conjugated linear peptide corresponding to human PKC phosphorylated at Ser729.

Application

Anti-phospho-PKCepsilon Antibody (Ser729) is a highly specific rabbit polyclonal antibody, that targets PKC epsilon & has been tested in western blotting.
Western Blotting/Peptide Inhibition Analysis: A 1:1,000 dilution from a representative lot detected phospho PKCepsilon (Ser729) in 10 µg of NIH/3T3 cell lysate.

Quality

Evaluated by Western Blotting/Peptide Inhibition Analysis in C6 cell lysate.

Western Blotting/Peptide Inhibition Analysis: A 1:1,000 dilution of this lot detected phospho PKCepsilon (Ser729) in 10 µg of C6 cell lysate.

Target description

~95 kDa and ~80 kDa observed. This protein has been observed at ~95 kDa and ~80 kDa (England, K. and Rumsby, M. G., et al. (2000). Biochem J. 352(Pt 1):19-26.).

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joana Santos et al.
Oncoscience, 2(5), 497-507 (2015-06-23)
Gastrin-releasing peptide receptor (GRPR) is known to be overexpressed in several human malignancies, including prostate cancer, and has been implicated in multiple important neoplastic signaling pathways. We recently have shown that GRPR is an ERG and ETV1 target gene in
Ioannis Smyrnias et al.
Journal of molecular and cellular cardiology, 117, 1-18 (2018-02-23)
The shortening of sarcomeres that co-ordinates the pump function of the heart is stimulated by electrically-mediated increases in [Ca2+]. This process of excitation-contraction coupling (ECC) is subject to modulation by neurohormonal mediators that tune the output of the heart to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service