Skip to Content
Merck

Skip To

06-554

Anti-STAT5B Antibody

Upstate®, from rabbit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
13
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human, mouse

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... STAT5B(6777)

General description

90-92kDa

Immunogen

amino acids 774-787 of human STAT5B (CGRPMDSQWIPHAQS)

Application

Anti-STAT5B Antibody is a Rabbit Polyclonal Antibody for detection of STAT5B also known as signal transducer & activator of transcription 5B & has been tested in WB.

Physical form

Format: Purified

Analysis Note

Control
Positive Antigen Control: Catalog #12-305, 3T3/A31 lysate. Add 2.5 μL of 2-mercapto-ethanol/100 μL of lysate and boil for 5 minutes to reduce the preparation. Load 20 μg of reduced lysate per lane for minigels.
routinely evaluated by immunoblot on mouse 3T3/A31 or human A431 cell lysates

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
07-135209-00707-140
biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

species reactivity

human, mouse

species reactivity

human

species reactivity

human

species reactivity

mouse

conjugate

unconjugated

conjugate

-

conjugate

-

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

technique(s)

western blot: suitable

technique(s)

dot blot: suitable, western blot: suitable

technique(s)

dot blot: suitable, western blot: suitable

technique(s)

electrophoretic mobility shift assay: suitable, immunocytochemistry: suitable, immunoprecipitation (IP): suitable, western blot: suitable


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters