Skip to Content
Merck
All Photos(1)

Key Documents

05-988

Sigma-Aldrich

Anti-Pras40 Antibody, Clone 73P21

clone 73P21, Upstate®, from mouse

Synonym(s):

40 kDa proline-rich AKT substrate, AKT1 substrate 1 (proline-rich), proline-rich Akt substrate, 40 kDa

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

73P21, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... AKT1S1(84335)

General description

PRAS40 (Proline Rich Akt Substrate, 40 kDa), also known as AKT1S1, is a ubiquitously express protein that is phosphorylated on residue Thr246 via the PI3K/Akt pathway. It was originally discovered as an Akt substrate as it has the putative Akt phosphorylation motif of RxRxxpS/pT2. This phosphorylation allows it to bind 14-3-3 and Raptor of the mTOR complex mTORC1. PRAS40 is known to bind to and regulate mTORC1 (mTOR, Raptor, mLST8) kinases activity that is activated by insulin downstream of PI3K and Akt and subsequently phosphorylates p70S6K and 4EBP1. PRAS40 is known to have a putative TOS motif (FVMDE) that allows it bind to Raptor of mTORC1 in the absence of insulin1. It is thought that through it binding to the Raptor, PRAS40 inhibits the kinase activity of mTORC1 by preventing to bind to its substrates such as p70S6K and 4EBP1.

Specificity

Recognizes PRAS40.
This antibody does not cross react with mouse.

Immunogen

Recombinant human PRAS40 (full-length) expressed in E. coli.

Application

Anti-Pras40 Antibody, Clone 73P21 is an antibody against Pras40 for use in IP & WB.
Immunoblot Analysis: A 01-1.0 μg/mL dilution of this lot detected PRAS40 in RIPA lysates of HeLa cells.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
PI3K, Akt, & mTOR Signaling

Quality

Routinely evaluated by western blot.

Western Blot Analysis:
A 0.1-1.0 µg/mL dilution of this lot detected PRAS40 in RIPA lysates of HeLa cells.

Target description

40 kDa

Physical form

Affinity purified
Format: Purified
Purified mouse monoclonal IgG1 in buffer containing PBS with 1% BSA and 0.1% sodium azide.

Storage and Stability

Stable for 1 years at -20°C from date of receipt.

Handling Recommendations:
Upon first thaw, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
RIPA lysates of HeLa cells.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David A Guertin et al.
Cancer cell, 15(2), 148-159 (2009-02-03)
mTOR complex 2 (mTORC2) contains the mammalian target of rapamycin (mTOR) kinase and the Rictor regulatory protein and phosphorylates Akt. Whether this function of mTORC2 is critical for cancer progression is unknown. Here, we show that transformed human prostate epithelial
Daniel W D West et al.
The American journal of clinical nutrition, 94(3), 795-803 (2011-07-29)
Ingestion of whey or casein yields divergent patterns of aminoacidemia that influence whole-body and skeletal muscle myofibrillar protein synthesis (MPS) after exercise. Direct comparisons of the effects of contrasting absorption rates exhibited by these proteins are confounded by their differing
Evaluating the therapeutic potential of mTOR inhibitors using mouse genetics.
Huawei Li,Jennifer L Cotton,David A Guertin
Methods in Molecular Biology null
Wenting Mi et al.
Oncotarget, 6(16), 13962-13977 (2015-05-12)
The mTORC1 inhibitors, rapamycin and its analogs, are known to show only modest antitumor activity in clinic, but the underlying mechanisms remain largely elusive. Here, we found that activated AKT signaling is associated with rapamycin resistance in breast and colon

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service