05-954
Anti-TCL1 Antibody
culture supernatant, Upstate®
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
biological source
mouse
Quality Level
antibody form
culture supernatant
antibody product type
primary antibodies
clone
monoclonal
species reactivity
human
manufacturer/tradename
Upstate®
technique(s)
immunohistochemistry: suitable
western blot: suitable
isotype
IgG
NCBI accession no.
UniProt accession no.
shipped in
dry ice
target post-translational modification
unmodified
Gene Information
human ... TCL1A(8115)
Specificity
TCL1A, no cross-reactivity with TML1 (TCL1B) or MTCP1
Immunogen
KLH-conjugated, synthetic peptide corresponding to amino acids 60-73 (RPMTPTQIGPSLLP) of human TCL1A
Application
Research Category
Inflammation & Immunology
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
Immunoglobulins & Immunology
This Anti-TCL1 Antibody is validated for use in WB, IH for the detection of TCL1.
Quality
Routinely evaluated by immunoblot in RIPA lysates from Raji cells
Target description
14 kD
Physical form
Cell culture supernatant
Storage and Stability
2 years at -20°C from date of shipment
Legal Information
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Structural basis for the co-activation of protein kinase B by T-cell leukemia-1 (TCL1) family proto-oncoproteins.
The Journal of Biological Chemistry, 279, 35890-35902 (2004)
TCL1: a new drug target in lymphoid and germ-cell malignancies?
The International Journal of Biochemistry & Cell Biology, 35, 1614-1618 (2003)
TCL1 is a diagnostic marker for intratubular germ cell neoplasia and classic seminoma.
Histopathology null
A systematic approach to diagnosis of mature T-cell leukemias reveals heterogeneity among WHO categories
Blood, 104, 328-335 (2004)
Crystal structures of Tcl1 family oncoproteins and their conserved surface features.
TheScientificWorldJournal, 2, 1876-1884 (2002)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service