Skip to Content
Merck
All Photos(1)

Documents

05-871

Sigma-Aldrich

Anti-Factor VIII Antibody, clone GMA-012

clone GMA-012, Upstate®, from mouse

Synonym(s):

Anti-Factor VIII Antibody, Clone GMA-012 Anti-Factor VIII, Factor VIII Detection Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

GMA-012, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

Specificity

Factor VIII

Immunogen

Purified human Factor VIII

Application

Detect Factor VIII using this Anti-Factor VIII Antibody, clone GMA-012 validated for use in ELISA & WB.
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Quality

routinely evaluated by immunoblot on human Factor VIII

Target description

Mr 280kDa

Physical form

Format: Purified
Protein G purified

Storage and Stability

stable 2 years at -20°C from date of shipment

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Coagulation factor VIII: structure and stability.
Wang, Wei, et al.
International Journal of Pharmaceutics, 259, 1-15 (2003)
Blood coagulation factor VIII: An overview.
Bhopale, G M and Nanda, R K
Journal of Biosciences, 28, 783-789 (2003)
Recombinant factor VIIa: review of efficacy, dosing regimens and safety in patients with congenital and acquired factor VIII or IX inhibitors.
Abshire, T and Kenet, G
Journal of Thrombosis and Haemostasis, 2, 899-909 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service