Skip to Content
Merck
All Photos(1)

Key Documents

05-837

Sigma-Aldrich

Anti-HFH-4/FOXJ1 Antibody, clone 3-19

ascites fluid, clone 3-19, Upstate®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

ascites fluid

antibody product type

primary antibodies

clone

3-19, monoclonal

species reactivity

rat, mouse, human

manufacturer/tradename

Upstate®

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... FOXJ1(2302)

Specificity

HFH-4/FOXJ1

Immunogen

Full length rat hepatocyte nuclear factor homolog (HFH-4)/forkhead box protein J1 (FOXJ1) GST fusion protein

Application

Anti-HFH-4/FOXJ1 Antibody, clone 3-19 is a high quality Mouse Monoclonal Antibody for the detection of HFH-4/FOXJ1 & has been validated in WB & IHC.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

routinely evaluated by immunoblot on modified RIPA lysates from rat lung

Target description

Mr 60kDa

Physical form

Ascites

Storage and Stability

Stable for 2 years at -20°C from date of shipment

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Engineering of fibrillar decorin matrices for a tissue-engineered trachea.
Svenja Hinderer,Marianne Schesny,Alexandra Bayrak,Bettina Ibold,Martina Hampel et al.
Biomaterials null
J W Tichelaar et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 47(6), 823-832 (1999-05-20)
HNF-3/forkhead homologue-4 (HFH-4), a transcription factor of the winged-helix/forkhead family, was detected by immunohistochemistry in tissue of the developing mouse. HFH-4 protein was present in epithelial cells of the lung, trachea, oviduct, and embryonic esophagus, and in ependymal cells lining
Tsung-Wei Huang et al.
Artificial cells, nanomedicine, and biotechnology, 47(1), 564-570 (2019-03-13)
Developing a biomaterial that promotes regeneration of both respiratory epithelium (RE) and olfactory neuroepithelium (ON) improves the surgical outcome of endoscopic sinus surgery. Although chitosan (CS) inhibits mucociliary differentiation of RE, it has been reported to regenerate ON. In addition

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service