Skip to Content
Merck
All Photos(1)

Key Documents

05-645

Sigma-Aldrich

Anti-phospho-HSP27 (Ser78) Antibody, clone JBW502

clone JBW502, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
JBW502, monoclonal
application:
WB
species reactivity:
human
technique(s):
western blot: suitable
citations:
4
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

JBW502, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pSer78)

Gene Information

human ... HSPB2(3316)

Specificity

phosphorylated HSP27 (Ser78)

Immunogen

peptide corresponding to amino acids 73-83 of human HSP27 with a C-terminus cysteine added for conjugation (YSRAL[pS]RQLSS-C)

Application

Anti-phospho-HSP27 (Ser78) Antibody, clone JBW502 is an antibody against phospho-HSP27 (Ser78) for use in WB.
Research Category
Protein Trafficking
Research Sub Category
Chaperones

Quality

routinely evaluated by immunoblot on phosphorylated HSP27 in 20mg of a cell lysate prepared from MCF-7 cells that were heat shocked for 30 minutes at 42°C, cultured for an additional 16 hours at 37°C, and then stimulated with 200mM arsenite for 30 minutes

Target description

27kDa

Linkage

Replaces: 04-447

Physical form

0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Format: Purified
Protein G Chromatography

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Heat shock protein 27 association with the I kappa B kinase complex regulates tumor necrosis factor alpha-induced NF-kappa B activation.
Park, KJ; Gaynor, RB; Kwak, YT
The Journal of Biological Chemistry null
Phosphorylation of Ser78 of Hsp27 correlated with HER-2/neu status and lymph node positivity in breast cancer.
Zhang, D; Wong, LL; Koay, ES
Molecular Cancer null
Yusuke Tamura et al.
Molecular oncology, 15(5), 1289-1307 (2020-12-21)
Development of innovative therapeutic modalities would address an unmet clinical need in the treatment of triple negative breast cancer (TNBC). Activation of retinoic acid-inducible gene-I (RIG-I)-like receptors (RLRs) such as melanoma differentiation-associated gene 5 (MDA5) and RIG-I in cancer cells
Eugene Chang et al.
Blood, 117(8), 2527-2537 (2010-12-07)
Actin filament remodeling regulates several endothelial cell (EC) processes such as contraction, migration, adhesion, and shape determination. Mitogen-activated protein kinase (MAPK)-activated protein kinase 2 (MK2)-mediated phosphorylation of heat-shock protein 27 kDa (HSP27) promotes actin filament remodeling, but little is known

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service