Skip to Content
Merck
All Photos(4)

Key Documents

05-593

Sigma-Aldrich

Anti-α-Dystroglycan Antibody, clone IIH6C4

ascites fluid, clone IIH6C4, Upstate®

Synonym(s):

Dystrophin-associated glycoprotein 1, dystroglycan 1, dystroglycan 1 (dystrophin-associated glycoprotein 1), dystrophin-associated glycoprotein-1, LARGE-glycan, Large glycan

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

antibody form

ascites fluid

clone

IIH6C4, monoclonal

species reactivity

human, mouse, canine, rat, guinea pig, rabbit

manufacturer/tradename

Upstate®

technique(s)

immunofluorescence: suitable
immunohistochemistry: suitable
inhibition assay: suitable
western blot: suitable

isotype

IgM

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... DAG1(1605)

General description

Dystroglycans are essential elements of the neuromuscular junction (NMJ). The gene for dystroglycan is expressed as a precursor protein that is post translationally cleaved into a 156 kDa extracellular peripheral membrane protein called alpha dystroglycan and a 43 kDa transmembrane protein, beta dystroglycan. The latter protein contains a PPxY motif that promotes binding to WW domain containing proteins, such as utrophin and dystrophin. Phosphorylation at tyrosine 892 within the PPxY motif may regulate c Src interactions with beta dystroglycan, as well as inhibit interactions with WW domain proteins. In skeletal muscle, beta dystroglycan is normally localized to the plasma membrane, however phosphorylation of Tyr892 leads to localization of beta dystroglycan to endosomal compartments along with c Src. Thus, phosphorylation at Tyr892 may have important roles in altering the localization of beta dystroglycan during NMJ formation.
This product may be used for research purposes only. Diagnostic use of this product requires a license from the University of Iowa Research Foundation, 214 Technology Innovation Center, Iowa City, IA 52242

Specificity

This antibody recognizes α-Dystroglycan/LARGE-glycan, Mr 156 kDa.

Immunogen

Rabbit skeletal muscle membrane preparation. Clone IIH6C4.

Application

Inhibition of Laminin Binding to Dystroglycan: An independent lab has shown, in a nitrocellulose overlay experiment, that this antibody inhibits binding of 125I-laminin to dystroglycan (Ervasti, J., et al. (1993).

Western Blot Analysis: A previous lot of this antibody was used on mouse muscle tissue lysate 3 months after shRNA induction, and in littermate controls (ctrl) and LARGE-null negative control (myd) (Goddeeris, M., et al. 2013, Nature).

Immunofluorescence: A previous lot of this antibody was used to detect α-Dystroglycan /LARGE-glycan in mouse muscle tissue (Goddeeris, M., et al. 2013, Nature).
Research Category
Metabolism
Research Sub Category
Muscle Physiology
This Anti-α-Dystroglycan Antibody, clone IIH6C4 is validated for use in IH, FUNC, WB for the detection of α-Dystroglycan.

Quality

Routinely evaluated by western blot on rabbit skeletal muscle.

Western Blot Analysis:
A 1:1000-1:2000 dilution of this lot detected α-Dystroglycan/LARGE-glycan in rabbit skeletal muscle.
Note: The use of WGA purified protein results in significantly cleaner blots and immunoprecipitates.
Post-translational modification of dystroglycan causes band broadening.

Target description

156 kDa

Physical form

Mouse ascites, in PBS containing 0.05% sodium azide and 30% glycerol.
Liquid at -20ºC.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt. For maximum recovery of product, centrifuge the vial prior to removing the cap.

Analysis Note

Control
Rabbit skeletal muscle lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Wu et al.
Gene therapy, 21(9), 785-793 (2014-06-20)
Antisense therapy with both chemistries of phosphorodiamidate morpholino oligomers (PMOs) and 2'-O-methyl phosphorothioate has demonstrated the capability to induce dystrophin expression in Duchenne muscular dystrophy (DMD) patients in phase II-III clinical trials with benefit in muscle functions. However, potential of
Membrane organization of the dystrophin-glycoprotein complex.
Ervasti, J M and Campbell, K P
Cell, 66, 1121-1131 (1991)
Huaiyu Hu et al.
Genes, 7(12) (2016-12-06)
Patients with type II lissencephaly, a neuronal migration disorder with ectopic neurons, suffer from severe mental retardation, including learning deficits. There is no effective therapy to prevent or correct the formation of neuronal ectopia, which is presumed to cause cognitive
Jihee Kim et al.
Skeletal muscle, 6, 3-3 (2016-02-24)
The defective glycosylation of α-dystroglycan is associated with a group of muscular dystrophies that are collectively referred to as the secondary dystroglycanopathies. Mutations in the gene encoding fukutin-related protein (FKRP) are one of the most common causes of secondary dystroglycanopathy
Katharine M Sharpe et al.
Journal of muscle research and cell motility, 34(5-6), 395-405 (2013-10-08)
Heart disease is a leading cause of death in patients with Duchenne muscular dystrophy (DMD). Patients with DMD lack the protein dystrophin, which is widely expressed in striated muscle. In skeletal muscle, the loss of dystrophin results in dramatically decreased

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service