Skip to Content
Merck
All Photos(1)

Key Documents

05-372

Sigma-Aldrich

Anti-GRB2 Antibody, clone 3F2

clone 3F2, Upstate®, from mouse

Synonym(s):

Anti-ASH, Anti-EGFRBP-GRB2, Anti-Grb3-3, Anti-MST084, Anti-MSTP084, Anti-NCKAP2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

3F2, monoclonal

species reactivity

bovine, mouse, hamster, rat, human

manufacturer/tradename

Upstate®

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GRB2(2885)

Specificity

N-terminal SH3 region of GRB2

Immunogen

Recombinant GRB2/Ash

Application

This Anti-GRB2 Antibody, clone 3F2 is validated for use in IH, WB for the detection of GRB2.

Quality

routinely evaluated in immunoblot on RIPA lysates of non-stimulated human A431 carcinoma cells, RIPA lysates of mouse 3T3/A31 fibroblasts, rat L6 myoblasts, Chinese hamster ovary cells or bovine brain extract

Target description

23-25kDa

Physical form

Format: Purified
PBS containing 50% glycerol

Analysis Note

Control
Positive Antigen Control: Catalog #12-301, non-stimulated A431 cell lysate. Add 2.5µL of 2-mercaptoethanol/100µL of lysate and boil for 5 minutes to reduce the preparation. Load 20µg of reduced lysate per lane for minigels.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Rozakis-Adcock et al.
Nature, 363(6424), 83-85 (1993-05-06)
Many tyrosine kinases, including the receptors for hormones such as epidermal growth factor (EGF), nerve growth factor and insulin, transmit intracellular signals through Ras proteins. Ligand binding to such receptors stimulates Ras guanine-nucleotide-exchange activity and increases the level of GTP-bound
c-Cbl is transiently tyrosine-phosphorylated, ubiquitinated, and membrane-targeted following CSF-1 stimulation of macrophages
Wang, Y., et al
The Journal of Biological Chemistry, 271, 17-20 (1996)
Protein-tyrosine phosphatase 1D modulates its own state of tyrosine phosphorylation
Stein-Gerlach, M., et al
The Journal of Biological Chemistry, 270, 24635-24637 (1995)
Ramya Ganesan et al.
Journal of leukocyte biology, 103(5), 867-883 (2018-04-16)
The uptake of cholesterol carried by low-density lipoprotein (LDL) is tightly controlled in the body. Macrophages are not well suited to counteract the cellular consequences of excess cholesterol leading to their transformation into "foam cells," an early step in vascular
T cell receptor-induced phosphorylation of Sos requires activity of CD45, Lck, and protein kinase C, but not ERK.
Zhao, H, et al.
The Journal of Biological Chemistry, 272, 21625-21634 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service