Skip to Content
Merck
All Photos(12)

Key Documents

05-1307

Sigma-Aldrich

Anti-Ubiquitin Antibody, Lys48-Specific, clone Apu2, rabbit monoclonal

clone Apu2, from rabbit

Synonym(s):

Anti-HEL-S-50

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

purified antibody

antibody product type

primary antibodies

clone

Apu2, monoclonal

species reactivity

rabbit, mouse, human, rat

technique(s)

cell based assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... UBAP2(55833)

General description

DI-UBIQUITIN PROTEIN AVAILABLE!
Di-Ubiquitin (Lys48-linked) Recombinant Protein (17-10408)
Polyubiquitin chains linked through the Lys48 residue of ubiquitin are most commonly associated with proteins targeted for proteosomal degradation.

In contrast, polyubiquitin chains linked through the Lys63 residue of ubiquitin (Ub) are associated with nonproteolytic functions such as signal transduction. Lys63-linked polyUb chains connect components of NFκB signaling in a highly regulated manner, and genetic evidence indicates the involvement of Lys63-linked Ub chains in stress response and DNA repair. Research in yeast has suggested Lys63-linked Ub chains stimulate endocytosis.

Specificity

Based on sequence homology, broad species cross-reactivity is expected.
Recognizes ubiquitin linked at Lys48. Does not recognize ubiquitin linked at Lys63 or free ubiquitin.

Immunogen

Epitope: Lys48 ubiquitination site
Immunogen used was human di-ubiquitin linked at Lys48.

Application

Immunohistochemistry:
Ubiquitin (Lys 48-specific) staining of rat brain sections (left figure) or human breast carcinoma (right figure). Staining pattern detects axons of neurons in brain tissue but not surrounding glial cells, and breast tumor cell cytoplasm is strongly detected without nuclear involvement.

Immunocytochemistry:
Confocal IF analysis of NIH/3T3, HeLa, and A431 cells using anti-Ubiquitin (Lys48 specific) (Red). Actin filaments were labeled with AlexaFluor 488-Phalloidin (Green). Nuclear material is stained with DAPI (Blue).

Flow cytometry:
Flow cytometry analysis of Jurkat cells using Anti-ubiquitin Lys48-specific (#05-1307) (blue) and rabbit IgG control (red).

Immunoprecipitation:
Rabbit monoclonal anti-ubiquitin (Lys-48-specific), clone Apu2, precipitated ubiquitin-containing proteins from HeLa cell lysates.

Inhibition Assay:
Preincubation of anti-ubiquitin, Lys48-specific, with K48-multiubiquitin chains inhibited binding to HeLa cell lysate in Western blots.
Anti-Ubiquitin Antibody, Lys48-Specific, clone Apu2 is a high quality Rabbit Monoclonal Antibody for the detection of Ubiquitin & has been validated in FC, ICC, IP, IHC, WB, PIA.
Research Category
Signaling
Research Sub Category
Ubiquitin & Ubiquitin Metabolism

Quality

Western Blot Analysis:
A 1:1000-1:5000 dilution of this lot detected Lys-48 ubiquitin chains by Western blotting.

Target description

varies

Physical form

Format: Purified
Protein A purified
Protein A purified recombinant purified recombinant rabbit monoclonal antibody that consists of a humanized fAb fragment fused to a rabbit Fc tail produced in a CHO recombinant antibody system. Supplied in 0.1M Tris-Glycine (pH7.4) 150 mM NaCl, 0.05% sodium azide

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.
For maximum recovery of product, centrifuge the vial prior to removing the cap.

Analysis Note

Control
Lys48 polyubiquitin chains.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Atypical ubiquitin chains: new molecular signals. 'Protein Modifications: Beyond the Usual Suspects' review series.
Ikeda, Fumiyo and Dikic, Ivan
EMBO Reports, 9, 536-542 (2008)
Lina Herhaus et al.
Nature communications, 4, 2519-2519 (2013-09-28)
SMAD transcription factors are key intracellular transducers of TGFβ cytokines. SMADs are tightly regulated to ensure balanced cellular responses to TGFβ signals. Ubiquitylation has a key role in regulating SMAD stability and activity. Several E3 ubiquitin ligases that regulate the
R Travis Taylor et al.
Methods (San Diego, Calif.), 55(2), 166-171 (2011-08-23)
Ubiquitin (Ub) conjugation to a substrate protein is a widely used cellular mechanism for control of protein stability and function, modulation of signal transduction pathways and antiviral responses. Identification and characterization of ubiquitinated viral proteins is an important step in
Grant S Stewart et al.
Cell, 136(3), 420-434 (2009-02-11)
The biological response to DNA double-strand breaks acts to preserve genome integrity. Individuals bearing inactivating mutations in components of this response exhibit clinical symptoms that include cellular radiosensitivity, immunodeficiency, and cancer predisposition. The archetype for such disorders is Ataxia-Telangiectasia caused
Keiko Taguchi et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(34), 13561-13566 (2012-08-09)
The Kelch-like ECH-associated protein 1 (Keap1)-NF-E2-related factor 2 (Nrf2) system is essential for cytoprotection against oxidative and electrophilic insults. Under unstressed conditions, Keap1 serves as an adaptor for ubiquitin E3 ligase and promotes proteasomal degradation of Nrf2, but Nrf2 is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service