Skip to Content
Merck

Skip To

04-771

Anti-Akt2/PKBβ Antibody, clone AW114, rabbit monoclonal

clone AW114, from rabbit

Synonym(s):

AKT2 kinase, Murine thymoma viral (v-akt) homolog-2, PKB beta, Protein kinase Akt-2, Protein kinase B, beta, rac protein kinase beta, v-akt murine thymoma viral oncogene homolog 2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
AW114, monoclonal
Application:
IP, WB
Citations:
-
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

AW114, monoclonal

species reactivity

rat, human, mouse

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... AKT2(208)

General description

Akt is the major known effector of the PI3 Kinase pathway. Generation of PIP3 results in the activation of PDK1, which phosphorylates Akt on Thr308, and another kinase complex, thought to be the mTORC2 complex (or possibly an intermediate kinase linking the two) that phosphorylates Akt on Ser473. These phosphorylations additively activate Akt Ser/Thr kinase activity, and the use of phosphorylation state-specific antibodies directed against either of these sites can imply Akt activation. Activation of Akt can be measured directly by immunoprecipitation followed by phosphorylation of a known substrate with radiolabeled ATP. Akt has been shown to phosphorylate over 70 substrates including TSC1&2 in the mTOR pathway, Bad on Ser136, GSK3 (which is inactivated by this phosphorylation), and the FoxO family of transcription factors (PRAS40, AS160).
~60 kDa

Immunogen

Epitope: a.a. 455-469
KLH-conjugated, synthetic peptide (CRYDSLGSLELDQRTH) corresponding to amino acids 455-469 of human Akt2/PKBβ, with an N-terminal cysteine added for conjugation purposes.

Application

Anti-Akt2/PKBβ Antibody, clone AW114 detects level of Akt2/PKBβ & has been published & validated for use in WB & IP.
Research Category
Signaling
Research Sub Category
PI3K, Akt, & mTOR Signaling

Biochem/physiol Actions

Predicted to cross-react with mouse.
Recognizes Akt2/PKBβ, Mr ~60 kDa.

Physical form

Cultured supernantant containing 0.05% sodium azide.
Format: Purified

Preparation Note

Stable for 1 year at -20ºC from date of receipt.
Handling Recommendations: Upon receipt, and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Control
L6 cell lysate
Western Blot Analysis: A 1:500 dilution of this antibody detected Akt2/PKBβ in HepG2 lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Replaces: 04-406

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
05-66904-79607-1398
clone

AW114, monoclonal

clone

11E6, monoclonal

clone

AW24, monoclonal

clone

polyclonal

biological source

rabbit

biological source

mouse

biological source

rabbit

biological source

rabbit

species reactivity

rat, human, mouse

species reactivity

rat, canine, mouse, human

species reactivity

human, rat, mouse

species reactivity

mouse, canine

conjugate

unconjugated

conjugate

-

conjugate

unconjugated

conjugate

-

Gene Information

human ... AKT2(208)

Gene Information

dog ... Akt1(490878)
human ... AKT1(207)

Gene Information

human ... AKT1(207)

Gene Information

bovine ... Akt1(280991)
human ... AKT1(207)
rat ... Akt1S1(292887)

antibody form

culture supernatant

antibody form

purified antibody

antibody form

culture supernatant

antibody form

affinity isolated antibody


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library






Global Trade Item Number

SKUGTIN
04-77104053252589157

Questions

Reviews

No rating value

Active Filters